Skip to Content
Merck
All Photos(1)

Key Documents

S0930000

Sodium valproate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Valproic acid sodium salt, 2-propylpentanoic acid sodium, Sodium 2-propylpentanoate, Sodium valproate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NaO2
CAS Number:
Molecular Weight:
166.19
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sodium valproate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].CCCC(CCC)C([O-])=O

InChI

1S/C8H16O2.Na/c1-3-5-7(6-4-2)8(9)10;/h7H,3-6H2,1-2H3,(H,9,10);/q;+1/p-1

InChI key

AEQFSUDEHCCHBT-UHFFFAOYSA-M

Gene Information

human ... ALDH5A1(7915)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium valproate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Anti-convulsant that also has efficacy as a mood stabilizer in bipolar disorder.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristen A Wellmann et al.
Brain research bulletin, 108, 1-9 (2014-08-19)
Prenatal exposure to valproic acid (VPA) alters rodent social interactions in a dose-dependent way: exposure to a high dose of VPA (>500 mg/kg) mid-gestation decreases social interactions whereas a moderate dose of VPA (350 mg/kg) increases peer-directed social behavior. The
Péter Pelsőczi et al.
Frontiers in behavioral neuroscience, 13, 295-295 (2020-02-06)
Autism spectrum disorder (ASD) is characterized by impaired socio-communicational function, repetitive and restricted behaviors. Valproic acid (VPA) was reported to increase the prevalence of ASD in humans as a consequence of its use during pregnancy. VPA treatment also induces autistic-like
Pengfei Shi et al.
BMC cancer, 14, 370-370 (2014-06-03)
Valproic acid (VPA), a histone deacetylase (HDAC) inhibitor, is reported to exert anti-tumor effects by upregulating the expression of the natural killer group 2D (NKG2D) ligands on tumor cells; however, the mechanisms vary in different tumor types, and the effect
Peter Štefánik et al.
Pharmacology, biochemistry, and behavior, 131, 42-50 (2015-02-11)
Autism is a neurodevelopmental disorder characterised by the disruption of social interactions. Autistic animal models play a crucial role in neurophysiologic research on this disorder. One of these models is based on rats that have been prenatally treated with valproic
Osvaldo Garrido-Acosta et al.
Journal of pharmacological and toxicological methods, 70(1), 66-69 (2014-05-27)
We determined the median effective dose (ED50) values for the anticonvulsants phenobarbital and sodium valproate using a modification of Lorke's method. This modification allowed appropriate statistical analysis and the use of a smaller number of mice per compound tested. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service