Skip to Content
Merck
All Photos(2)

Key Documents

SML0061

Sigma-Aldrich

SAHA

≥98% (HPLC), powder, HDAC inhibitor

Synonym(s):

N-hydroxy-N′-phenyl-octanediamide, Suberoylanilide hydroxamic acid, Vorinostat

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O3
CAS Number:
Molecular Weight:
264.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

SAHA, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥15 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

ONC(=O)CCCCCCC(=O)Nc1ccccc1

InChI

1S/C14H20N2O3/c17-13(15-12-8-4-3-5-9-12)10-6-1-2-7-11-14(18)16-19/h3-5,8-9,19H,1-2,6-7,10-11H2,(H,15,17)(H,16,18)

InChI key

WAEXFXRVDQXREF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

SAHA may be used to study gene regulation, transcription regulation and cell signaling.

Biochem/physiol Actions

SAHA or Vorinostat facilitates the transcription of genes that result in apoptosis, differentiation and growth arrest. It has been observed to give beneficial results in lymphoma but not in solid tumors.
Vorinostat or suberoylanilide hydroxamic acid (SAHA) is a potent, reversible pan-histone deacetylase (HDAC) inhibitor. It inhibits both class I and class II HDACs, altering gene transcription and inducing cell cycle arrest and/or apoptosis in a wide variety of transformed cells.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert W Robey et al.
Molecular pharmaceutics, 8(6), 2021-2031 (2011-09-09)
The histone deacetylase inhibitors (HDIs) have shown promise in the treatment of a number of hematologic malignancies, leading to the approval of vorinostat and romidepsin for the treatment of cutaneous T-cell lymphoma and romidepsin for the treatment of peripheral T-cell
Sarah W Gordon et al.
American journal of clinical oncology, 42(8), 649-654 (2019-07-16)
Preclinical data suggest histone deacetylase inhibitors improve the therapeutic index of sorafenib. A phase I study was initiated to establish the recommended phase 2 dose of sorafenib combined with vorinostat in patients with unresectable hepatocellular carcinoma. Patients received vorinostat (200
Gloria Manzotti et al.
Journal of experimental & clinical cancer research : CR, 38(1), 346-346 (2019-08-10)
RUNX2 is a Runt-related transcription factor required during embryogenesis for skeletal development and morphogenesis of other organs including thyroid and breast gland. Consistent evidence indicates that RUNX2 expression is aberrantly reactivated in cancer and supports tumor progression. The mechanisms leading
Sachith Mettananda et al.
Scientific reports, 9(1), 11649-11649 (2019-08-14)
β-Thalassaemia is one of the most common monogenic diseases with no effective cure in the majority of patients. Unbalanced production of α-globin in the presence of defective synthesis of β-globin is the primary mechanism for anaemia in β-thalassaemia. Clinical genetic
Weiwei Yu et al.
British journal of pharmacology, 174(20), 3608-3622 (2017-07-28)
Intrinsic and/or acquired resistance of epidermal growth factor receptor (EGFR) tyrosine kinase inhibitors (TKIs) commonly occurs in patients with non-small-cell lung cancer (NSCLC). Here, we developed a combined therapy of histone deacetylase inhibition by a novel HDAC inhibitor, YF454A, with

Articles

We presents an article on Autophagy in Cancer Promotes Therapeutic Resistance

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service