Skip to Content
Merck
All Photos(2)

Key Documents

87613

Supelco

Erythrosin B

analytical standard

Synonym(s):

Erythrosin extra bluish, 2′,4′,5′,7′-Tetraiodofluorescein disodium salt, Acid Red 51, Iodoeosin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6I4Na2O5
CAS Number:
Molecular Weight:
879.86
Colour Index Number:
45430
Beilstein:
1443945
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E127
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].[O-]c1c(I)cc2c(Oc3c(I)c([O-])c(I)cc3C24OC(=O)c5ccccc45)c1I

InChI

1S/C20H8I4O5.2Na/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20;;/h1-6,25-26H;;/q;2*+1/p-2

InChI key

RAGZEDHHTPQLAI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Erythrosin B belongs to the class of xanthene dyes, which can find applications as a food colorant.(12} It can exhibit light absorption in the range of 500-550 nm, hence can lead to the formation of reactive oxygen species, which can inactivate the microorganisms, thereby acting like an antimicrobial agent.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

359.6 °F - closed cup

Flash Point(C)

182 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced antimicrobial effect of ultrasound by the food colorant Erythrosin B
Bastarrachea.JL, et al.
Food Research International, 100, 344-351 (2017)
Vagner Roberto Batistela et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 889-897 (2011-05-10)
Xanthenes form to an important class of dyes which are widely used. Most of them present three acid-base groups: two phenolic sites and one carboxylic site. Therefore, the pKa determination and the attribution of each group to the corresponding pKa
Chueh-Pin Chen et al.
Photochemistry and photobiology, 88(3), 570-576 (2012-01-31)
The growing resistance to antibiotics has rendered antimicrobial photodynamic inactivation (PDI) an attractive alternative treatment modality for infectious diseases. Chitosan (CS) was shown to further potentiate the PDI effect of photosensitizers and was therefore used in this study to investigate
Laura Carmen Apostol et al.
Biodegradation, 23(5), 725-737 (2012-03-23)
Biodegradation of a xanthene dyes was investigated for the first time using anaerobic granular sludge. On a first screening, biomass was able to decolorize, at different extents, six azo dye solutions: acid orange 7, direct black 19, direct blue 71
H Edward Wong et al.
PloS one, 6(10), e25752-e25752 (2011-10-15)
Alzheimer's disease (AD) is the most common form of dementia. AD is a degenerative brain disorder that causes problems with memory, thinking and behavior. It has been suggested that aggregation of amyloid-beta peptide (Aβ) is closely linked to the development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service