200964
Erythrosin B
Dye content ≥95 %
Synonym(s):
Solvent Red 140
About This Item
Recommended Products
form
powder
Quality Level
composition
Dye content, ≥95%
technique(s)
microbe id | staining: suitable
color
orange to red-orange
mp
303 °C (dec.) (lit.)
solubility
1 M NH4OH: 1 mg/mL
1 M NH4OH: soluble 1 mg/mL, orange to red
ε (extinction coefficient)
≥102000 at 526-534 nm at 0.005 g/L (95% Methanol 5% 0.1M Na2CO3)
≥13000 at 310-318 nm in 1 M NH4OH at 0.005 g/L (95% Methanol 5% 0.1M Na2CO3)
≥32000 at 258-266 nm at 0.005 g/L (95% Methanol 5% 0.1M Na2CO3)
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
Oc1c(I)cc2c(Oc3c(I)c(O)c(I)cc3C24OC(=O)c5ccccc45)c1I
InChI
1S/C20H8I4O5/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20/h1-6,25-26H
InChI key
OALHHIHQOFIMEF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
fluorescence lifetime
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service