Skip to Content
Merck
All Photos(2)

Key Documents

68724

Supelco

Fast Green FCF

analytical standard

Synonym(s):

Food green 3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C37H34N2O10S3Na2
CAS Number:
Molecular Weight:
808.85
Colour Index Number:
42053
Beilstein:
5718212
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E143
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

290 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].CCN(Cc1cccc(c1)S([O-])(=O)=O)c2ccc(cc2)C(=C3/C=CC(\C=C3)=[N+](\CC)Cc4cccc(c4)S([O-])(=O)=O)\c5ccc(O)cc5S([O-])(=O)=O

InChI

1S/C37H36N2O10S3.2Na/c1-3-38(24-26-7-5-9-33(21-26)50(41,42)43)30-15-11-28(12-16-30)37(35-20-19-32(40)23-36(35)52(47,48)49)29-13-17-31(18-14-29)39(4-2)25-27-8-6-10-34(22-27)51(44,45)46;;/h5-23H,3-4,24-25H2,1-2H3,(H3,41,42,43,44,45,46,47,48,49);;/q;2*+1/p-2

InChI key

RZSYLLSAWYUBPE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Fast Green FCF is a synthetic organic dye, which belongs to the triphenylmethane group. It can be found in a wide range of applications in food and beverage industry. It can also be used in scientific research for the staining of proteins and identification of extracellular recording or injection sites in electrophysiological experiments.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Fast Green FCF dye is used for protein staining in IEF, native PAGE and SDS-PAGE. Fast Green staining is linear over a wider range of protein concentrations than Brilliant Blue R. After electrophoresis, fixing the proteins in the gel is recommended for maximum sensitivity. The gel can then be stained for 2 hr with 0.1% Fast Green FCF in either 30% (v/v) ethanol, 10% (v/v) acetic acid or in 7% acetic acid. The gel can be destained with either 30% (v/v) ethanol, 10% (v/v) acetic acid, or 7% acetic acid. The stained gel can be scanned at 625 nm. The detection sensitivity is approx. 30% that with Brilliant Blue R.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fast Green FCF (Food Green 3) inhibits synaptic activity in rat hippocampal interneurons
Hooft V.AJ
Neuroscience Letters, 318, 163-165 (2002)
Nonlinear responses and optical limiting behavior of fast green FCF dye under a low power CW He?Ne laser irradiation
Jamshidi-Ghaleh K, et al.
Optics Communications, 271, 551-554 (2007)
D Wittekind et al.
Analytical and quantitative cytology, 4(4), 286-294 (1982-12-01)
The morphologic and spectroscopic characteristics of a new and reproducible modification of the Papanicolaou stain are briefly described. The main features of this modification are (1) replacement of the natural dye hematoxylin by the synthetic and chemically well defined dye
Sumiko Tsuji et al.
Journal of chromatography. A, 1101(1-2), 214-221 (2005-10-26)
The existence of five of the six expected isomers in commercial Fast Green FCF (G3: Food Green No. 3, FD&C Green No. 3, CAS No. 2353-45-9, C.I. No. 42053), the main product of which is m,m-G3 and the sub-products of
Alok Mittal et al.
Journal of hazardous materials, 163(2-3), 568-577 (2008-08-12)
De-Oiled Soya, an agricultural waste material and Bottom Ash a waste of power plants, have been used as adsorbents for the removal and recovery of a triarylmethane dye Fast Green FCF from wastewater. Batch studies have been carried by observing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service