Skip to Content
Merck
All Photos(3)

Key Documents

860867

Sigma-Aldrich

Brilliant Cresyl Blue ALD

Certified by the Biological Stain Commission

Synonym(s):

3-Amino-7-(diethylamino)-2-methylphenoxazin-5-ium (T-4)-tetrachlorozincate (2:1), Brilliant Blue C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20ClN3O · 0.5ZnCl2
CAS Number:
Molecular Weight:
385.96
Beilstein:
5690713
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Quality Level

form

powder or crystals

composition

Dye content, ≥50%

technique(s)

microbe id | staining: suitable

mp

233-236 °C (lit.)

λmax

622 nm

ε (extinction coefficient)

≥20000 at 237-241 nm in 50% ethanol at 0.005 g/L
≥20000 at 248-252 nm in 50% ethanol at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.CC\[N+](CC)=C1\C=CC2=Nc3cc(C)c(N)cc3OC2=C1.CC\[N+](CC)=C4\C=CC5=Nc6cc(C)c(N)cc6OC5=C4

InChI

1S/2C17H20N3O.4ClH.Zn/c2*1-4-20(5-2)12-6-7-14-16(9-12)21-17-10-13(18)11(3)8-15(17)19-14;;;;;/h2*6-10H,4-5,18H2,1-3H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

SIHASHFMDFXGSX-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Brilliant cresyl blue ALD has been used for the staining of reticulocytes.[1]

Biochem/physiol Actions

Brilliant cresyl blue ALD is responsible for the staining of RNA in reticulocytes.[2]

Other Notes

The structure of Brilliant Cresyl Blue ALD contains a 2-methyl modification on the phenoxazine core, while the structure of the historic Brilliant Cresyl Blue contains a 8-methyl modification on the phenoxazine core. The biological staining application is the same for both structures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammatory and oxidative stress phenotypes in transgenic sickle cell mice.
Charrin E, et al.
Blood Cells, Molecules and Diseases, 62, 13-13 (2016)
Pal GK and Pal P
Textbook Of Practical Physiology (2006)
Clark, G., Ed.
Staining Procedures, 176-176 (1981)
A Zh Baltabekova et al.
Molecular biotechnology, 57(8), 746-755 (2015-04-04)
Immunoglobulin E (IgE) plays a central role in type I hypersensitivity including allergy and asthma. Novel treatment strategy envisages development of a therapeutic vaccine designed to elicit autologous blocking antibodies against the IgE. We sought to develop an IgE-epitope antigen
David Berthier et al.
PloS one, 10(5), e0126498-e0126498 (2015-05-09)
Animal African Trypanosomosis particularly affects cattle and dramatically impairs livestock development in sub-Saharan Africa. African Zebu (AFZ) or European taurine breeds usually die of the disease in the absence of treatment, whereas West African taurine breeds (AFT), considered trypanotolerant, are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service