Skip to Content
Merck
All Photos(1)

Key Documents

16035

Sigma-Aldrich

Brilliant Cresyl blue solution

for microscopy, 0.3 % (w/v) in ethanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
6035110
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

grade

for microscopy

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

composition

dye ( C.I. 51010), 0.3 % (w/v) (in ethanol)

concentration

0.3 % (w/v) in ethanol

density

0.808 g/mL at 20 °C

εmax

6.0 in 50% ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC1=C(N)C=C([O+]=C(C=C(N(CC)CC)C=C2)C2=N3)C3=C1.Cl[Zn-2](Cl)(C)(Cl)Cl.CC4=C(N)C=C([O+]=C(C=C(N(CC)CC)C=C5)C5=N6)C6=C4

InChI

1S/2C17H20N3O.4ClH.Zn/c2*1-4-20(5-2)12-6-7-14-16(9-12)21-17-10-13(18)11(3)8-15(17)19-14;;;;;/h2*6-10H,4-5,18H2,1-3H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

SIHASHFMDFXGSX-UHFFFAOYSA-J

General description

Brilliant Cresyl blue is a phenoxazine dye.

Application

Brilliant Cresyl blue may be used for staining Hienz bodies, in an experimental study to investigate the importance of peroxiredoxin II, for sustaining life span of erythrocytes in mice.

Other Notes

Staining of immature red cells and platelets

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service