Skip to Content
Merck
All Photos(3)

Key Documents

318922

Sigma-Aldrich

Ferroin indicator solution

0.1 wt. % in H2O

Synonym(s):

1,10-Phenanthroline iron(II) sulfate complex, o-Phenanthroline ferrous sulfate complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Fe(C12H8N2)3]SO4
CAS Number:
Molecular Weight:
692.52
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

liquid

Quality Level

concentration

0.1 wt. % in H2O

density

0.999 g/mL at 25 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Fe++].[O-]S([O-])(=O)=O.c1cnc2c(c1)ccc3cccnc23.c4cnc5c(c4)ccc6cccnc56.c7cnc8c(c7)ccc9cccnc89

InChI

1S/3C12H8N2.Fe.H2O4S/c3*1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;1-5(2,3)4/h3*1-8H;;(H2,1,2,3,4)/q;;;+2;/p-2

InChI key

CIWXFRVOSDNDJZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ferroin serves as an indicator of oxidation reduction reactions in titrimetric analysis. Ferroin is the complex of 1,10-phenanthroline and Fe(II). It is also used as a catalyst for Belousov−Zhabotinsky reaction.

Application

Ferroin indicator solution is suitable for wastewater analysis of the parameters like chemical oxygen demand (COD) and total organic carbon (TOC).

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Artificial Life Models in Hardware, 236-236 (2009)
Separation, Preconcentration and Spectrophotometry in Inorganic Analysis, 229-229 (2000)
Chemical Demonstrations: A Handbook for Teachers of Chemistry, 2, 261-261 (1985)
Luciano A Cardoso et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 151(3), 313-321 (2007-06-05)
Glutathione reductase (GR) carries out the enzymatic reduction of glutathione disulfide (GSSG) to its reduced form (GSH) at the expense of the reducing power of NADPH. Previous studies have shown that GR from several species is progressively inactivated in the
Laszló Hegedüs et al.
The journal of physical chemistry. A, 110(47), 12839-12844 (2006-11-28)
In the present mechanistic schemes of the ferroin-catalyzed oscillatory Belousov-Zhabotinsky (BZ) reaction the oxidation of the organic substrates (bromomalonic or malonic acid) by ferriin (the oxidized form of the catalyst) plays an important role. As the organic products of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service