Skip to Content
Merck
All Photos(1)

Key Documents

1.01384

Sigma-Aldrich

Brilliant cresyl blue solution

for the staining of reticulocytes for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.21

Quality Level

form

liquid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

pH

3.7 (20 °C in H2O)

density

1.01 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

The Brilliant cresyl blue solution - for the staining of reticulocytes - for microscopy, is a ready-to-use solution used for human-medical cell diagnosis and serves the hematological investigation of sample material of human origin.
The dye stains the nucleic acids in the reticulocytes (Substantia granulofilamentosa), thus enabling reticulocytes to be morphologically distinguished from erythrocytes. The Substantia granulofilamentosa (ribonucleoproteins) can be rendered visible with fresh, non-fixed, young erythrocytes in a supervital staining procedure. Characterized by an intense black-blue hue, it is possible both to determine the number of the reticulocytes (visual microscopic counting) and also to visualize their morphological aspects. Four stages of substantia granulofilamentosa maturation can be distinguished depending on the stage of reticulocyte development: coiled skein (I), incomplete network (II), complete network (III) and granular form (IV). In peripheral blood the development stages III and IV are found most commonly.
The 100 ml bottle provides ca. 1000 applications. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Application test (Reticulocyte staining): passes test

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service