Skip to Content
Merck
All Photos(1)

Key Documents

81942

Sigma-Aldrich

Propionic anhydride

purum, ≥96.0% (NT)

Synonym(s):

Propanoic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CO)2O
CAS Number:
Molecular Weight:
130.14
Beilstein:
507066
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.5 (vs air)

Quality Level

vapor pressure

10 mmHg ( 57.7 °C)

grade

purum

Assay

≥96.0% (NT)

form

liquid

autoignition temp.

545 °F

expl. lim.

11.9 %

refractive index

n20/D 1.404 (lit.)
n20/D 1.404

bp

167 °C (lit.)

mp

−43 °C (lit.)

solubility

H2O: decomposes (when in contact with water)

density

1.010 g/mL at 20 °C
1.015 g/mL at 25 °C (lit.)

functional group

anhydride

SMILES string

CCC(=O)OC(=O)CC

InChI

1S/C6H10O3/c1-3-5(7)9-6(8)4-2/h3-4H2,1-2H3

InChI key

WYVAMUWZEOHJOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Propionic anhydride can be used:
  • As an esterifying reagent in the modification of cassava starch by acylation reaction.
  • As a reactant along with anisole in the synthesis of 4-methoxypropiophenone using various solid acid catalysts via Friedel−Crafts acylation reaction.
  • As a reagent in the synthesis of cyanohydrin esters by treating KCN with aldehyde in presence of bimetallic titanium(salen) complex catalyst.

Other Notes

Sales restrictions may apply

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of cassava starch by using propionic anhydride and properties of the starch-blended polyester polyurethane
Santayanon R and Wootthikanokkhan J
Carbohydrate Polymers, 51(1), 17-24 (2003)
Synthetic and mechanistic studies on asymmetric cyanohydrin synthesis using a titanium (salen) bimetallic catalyst
Belokon YN, et al.
Tetrahedron, 60(46), 10433-10447 (2004)
Dimensional stabilisation and strength of particleboard by chemical modification with propionic anhydride.
Papadopoulos AN & Gkaraveli A
European Journal of Wood and Wood Products, 61(2), 142-144 (2003)
Robert Meatherall et al.
Journal of analytical toxicology, 33(8), 525-531 (2009-10-31)
Azide in human blood and plasma samples was derivatized with propionic anhydride in a headspace vial without prior sample preparation. The reaction proceeds quickly at room temperature to form propionyl azide. A portion of the headspace was assayed by gas
Surendra Nimesh et al.
International journal of pharmaceutics, 337(1-2), 265-274 (2007-01-27)
Polyethylenimine (750 kDa) has been derivatized to influence the proton sponge mechanism and hydrophobic-hydrophilic balance. The polymer was acylated using acid anhydrides of varying carbon chain length, followed by cross-linking with PEG-bis-P to form compact nanoparticles. The chemical linkages in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service