198218
Patent Blue VF
Dye content 50 %
Synonym(s):
Acid Blue 1, Sulfan Blue
About This Item
Recommended Products
form
powder
Quality Level
composition
Dye content, 50%
technique(s)
microbe id | staining: suitable
solubility
H2O: 1 mg/mL
λmax
635 nm (2nd)
-410 nm
ε (extinction coefficient)
≥50000 at 635-641 nm in H2O
≥5300 at 408-414 nm in H2O
≥7500 at 308-314 nm in H2O
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[Na+].CCN(CC)c1ccc(cc1)\C(=C2/C=CC(\C=C2)=[N+](\CC)CC)c3ccc(cc3S([O-])(=O)=O)S([O-])(=O)=O
InChI
1S/C27H32N2O6S2.Na/c1-5-28(6-2)22-13-9-20(10-14-22)27(21-11-15-23(16-12-21)29(7-3)8-4)25-18-17-24(36(30,31)32)19-26(25)37(33,34)35;/h9-19H,5-8H2,1-4H3,(H-,30,31,32,33,34,35);/q;+1/p-1
InChI key
SJEYSFABYSGQBG-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
- in combination with 99mTc phytate to detect sentinel lymph node (SLN) in early stage cervical cancer patients.
- to detect heat shock protein (HSP) gp96 induced draining lymph nodes (LNs) in mice.
- to detect the effect of Asn-Gly-Arg (NGR)-tumor necrosis factor (TNF) on tumor perfusion in murine model.
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Induces Maturation and Migration of
CD11c1 Cells In Vivo
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service