Skip to Content
Merck
All Photos(4)

Key Documents

402907

Sigma-Aldrich

Propionic acid

ACS reagent, ≥99.5%

Synonym(s):

Acid C3, Propanoic acid, Propanyl acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COOH
CAS Number:
Molecular Weight:
74.08
Beilstein:
506071
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021305
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

2.55 (vs air)

vapor pressure

2.4 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

955 °F

expl. lim.

12.1 %

analyte chemical class(es)

amino acids

impurities

≤0.002% Carbonyl compounds
≤0.10% Readily oxidizable substances (as HCOOH)
≤0.15% water

evapn. residue

≤0.01%

color

APHA: ≤20

refractive index

n20/D 1.386 (lit.)

bp

141 °C (lit.)

mp

−24-−23 °C (lit.)

solubility

H2O: soluble

density

0.993 g/mL at 25 °C (lit.)

cation traces

heavy metals (as Pb): ≤0.001%

functional group

carboxylic acid

SMILES string

CCC(O)=O

InChI

1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)

InChI key

XBDQKXXYIPTUBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propionic acid (PA) also known as propanoic acid is a short chain fatty acid mainly used as food preservative. It is one of the main metabolic end product formed during the fermentation of undigested food in the colon by the microbiota. Its manufacture by glycerol/glucose co-fermentation using Propionibacterium acidipropionici has been reported. Crystal structure study reveals that PA crystals are monoclinic with space group P21/c.
Propionic acid (PA) is a naturally occurring carboxylic acid, which in its pure state exists as a colorless corrosive liquid with an unpleasant odor. It is miscible in water. Industrially it is produced by hydrocarboxylation of ethylene in presence nickel carbonyl as a catalyst. PA has been found to reduce food intake, lower the fatty acids content in plasma and liver, might improve tissue insulin sensitivity and exerts immunosuppressive actions. Propionic acid is an excellent raw material as it is stable, cheap and safe and can even be used as a food additive.

Application

Propionic acid may be used to alter limestone in order to increase its ability of CO2 uptake.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yin Liu et al.
Current microbiology, 62(1), 152-158 (2010-06-15)
Cosubstrates fermentation is such an effective strategy for increasing subject metabolic products that it could be available and studied in propionic acid production, using glycerol and glucose as carbon resources. The effects of glycerol, glucose, and their mixtures on the
Derrick F MacFabe et al.
Behavioural brain research, 217(1), 47-54 (2010-10-13)
Recent evidence suggests that a variety of environmental factors, including dietary and gastrointestinal agents, may contribute to autism spectrum disorders (ASD). Here we administered propionic acid (PPA), a short chain fatty acid that is used as a food preservative and
The crystal structure of propionic acid.
Strieter FJ, et al.
Acta Crystallographica, 15(12), 1233-1239 (1962)
Enhancement of CO2 capture capacity by modifying limestone with propionic acid.
Sun R, et al.
Powder Technology, 233, 8-14 (2013)
Nucleophilic ?-Carbon Activation of Propionic Acid as a 3-Carbon Synthon by Carbene Organocatalysis
Jin, Zhichao, et al.
Chemistry?A European Journal , 21 (26), 9360-9363 (2015)

Articles

The detection and quantification of biomarkers are essential for medical diagnostics, environmental monitoring, and bioresearch.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service