Skip to Content
Merck
All Photos(1)

Key Documents

69471

Sigma-Aldrich

Trimethoxymethylsilane

purum, ≥98.0% (GC)

Synonym(s):

Methyltrimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3Si(OCH3)3
CAS Number:
Molecular Weight:
136.22
Beilstein:
1736151
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.371 (lit.)
n20/D 1.371

bp

102-104 °C (lit.)

density

0.955 g/mL at 25 °C (lit.)

SMILES string

CO[Si](C)(OC)OC

InChI

1S/C4H12O3Si/c1-5-8(4,6-2)7-3/h1-4H3

InChI key

BFXIKLCIZHOAAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F

Flash Point(C)

9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Gao et al.
Langmuir : the ACS journal of surfaces and colloids, 26(2), 1239-1246 (2009-09-03)
The immobilization of biological species such as proteins and enzymes in sol-gel hosts is currently an area of intense research activity. However, the majority of these studies have been directed toward investigating the biological activity or physicochemical properties of the
Sameer Kulkarni et al.
Journal of chromatography. A, 1174(1-2), 50-62 (2007-11-21)
Sol-gel coating with covalently bonded low-molecular-weight (MW<300 Da) poly(ethylene glycol) (PEG) chains was developed for capillary microextraction (CME). The sol-gel chemistry proved effective in the immobilization of low-molecular-weight PEGs thanks to the formation of chemical bonds between the organic-inorganic hybrid
Braden C Giordano et al.
Electrophoresis, 27(21), 4295-4302 (2006-10-06)
Microchip-based CEC of nitroaromatic and nitramine explosives with UV absorbance detection is described. The stationary phase was deposited in the microchip using the sol-gel process. Unique to this work, is the exclusive use of alkylated-trimethoxysilane precursors in the gel solution.
H El Rassy et al.
Chembiochem : a European journal of chemical biology, 4(2-3), 203-210 (2003-03-05)
Highly porous silica aerogels with differing balances of hydrophobic and hydrophilic functionalities were studied as a new immobilization medium for enzymes. Two types of lipases from Candida rugosa and Burkholderia cepacia were homogeneously dispersed in wet gel precursors before gelation.
Daniel A Riccio et al.
Biomaterials, 30(27), 4494-4502 (2009-06-09)
The synthesis, material characterization, and in vitro biocompatibility of S-nitrosothiol (RSNO)-modified xerogels are described. Thiol-functionalized xerogel films were formed by hydrolysis and co-condensation of 3-mercaptopropyltrimethoxysilane (MPTMS) and methyltrimethoxysilane (MTMOS) sol-gel precursors at varying concentrations. Subsequent thiol nitrosation via acidified nitrite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service