Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trimethoxymethylsilane
Synonym(s):
Methyltrimethoxysilane, Trimethoxymethylsilane
Linear Formula:
CH
3
Si(OCH
3
)
3
CAS Number:
1185-55-3
Molecular Weight:
136.22
EC Number:
214-685-0
Products
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Purum (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
246174
Trimethoxymethylsilane,
98%
Expand
View Pricing
440175
Trimethoxymethylsilane,
95%
Expand
View Pricing
8.14178
Trimethoxymethylsilane,
for synthesis
Expand
View Pricing
69471
Trimethoxymethylsilane,
purum, ≥98.0% (GC)
Expand
View Pricing