Skip to Content
Merck
All Photos(1)

Key Documents

246174

Sigma-Aldrich

Trimethoxymethylsilane

98%

Synonym(s):

Methyltrimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3Si(OCH3)3
CAS Number:
Molecular Weight:
136.22
Beilstein:
1736151
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.06

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.371 (lit.)

bp

102-104 °C (lit.)

density

0.955 g/mL at 25 °C (lit.)

SMILES string

CO[Si](C)(OC)OC

InChI

1S/C4H12O3Si/c1-5-8(4,6-2)7-3/h1-4H3

InChI key

BFXIKLCIZHOAAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethoxymethylsilane (MTM) is an organosilicon compound widely used as a precursor for the preparation of silica-based materials, which finds the applications in various fields. Particularly in molecular assembly, linking nano building blocks, and selective synthesis oligosiloxane compounds. It can also be utilized as a crosslinker in the synthesis of polysiloxane polymers.

Application

Trimethoxymethylsilane can be used:
  • As a silica source for synthesizing polyethyleneimine-silica (PEI-silica) organic-inorganic hybrid particles.
  • To transform hydrophilic ceramic surfaces to hydrophobic by modifying the -OH groups.
  • To modify silica aerogels by inducing hydrophobicity and enhancing mechanical properties without affecting transparency.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F

Flash Point(C)

9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Utilization of alkoxysilyl groups for the creation of structurally controlled siloxane-based nanomaterials
Kuroda K, et al.
Chemistry of Materials, 26(1), 211-220 (2014)
The formation of polyethyleneimine?trimethoxymethylsilane organic?inorganic hybrid particles.
Neville F, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 431, 42-50 (2013)
Organo-modified silica aerogels and implications for material hydrophobicity and mechanical properties.
Martin L, et al.
Journal of Materials Chemistry, 18(2), 207-213 (2008)
Investigation of Silane Modified Ceramic Surface of Porous Mullite Ceramics.
Markovska I, et al.
World Academy of Science, Engineering and Technology, 79(7), 272-276 (2013)
Casseday P Richers et al.
Inorganic chemistry, 55(12), 5744-5746 (2016-06-04)
The commercially practiced conversion of trimethoxymethylsilane (MTM) to [OSi(OMe)Me)]n polymers and resins is assumed to proceed via the silanol (MeO)2MeSiOH. Access to this crucial silanol is gained via the synthesis of (MeO)2MeSiONa, the first methoxysilanoate to be crystallographically characterized. Mild

Articles

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service