Skip to Content
Merck
All Photos(1)

Key Documents

175579

Sigma-Aldrich

Triethoxymethylsilane

99%

Synonym(s):

Methyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3Si(OC2H5)3
CAS Number:
Molecular Weight:
178.30
Beilstein:
1742453
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

vapor pressure

11 mmHg ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.383 (lit.)

bp

141-143 °C (lit.)

density

0.895 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](C)(OCC)OCC

InChI

1S/C7H18O3Si/c1-5-8-11(4,9-6-2)10-7-3/h5-7H2,1-4H3

InChI key

CPUDPFPXCZDNGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethoxymethylsilane is an organosilane that can be used to prepare:
  • An oxidizing catalyst known with excellent features such as selectivity, stability, and recyclability.
  • A hydrogel heterogeneous platinum catalyst applicable in the chemoselective hydrogenation of nitroarenes.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silia Cat TEMPO: An Effective and Useful Oxidizing Catalyst
Michaud A, et al.
Organic Process Research & Development, 11(4), 766-768 (2007)
A new class of heterogeneous platinum catalysts for the chemoselective hydrogenation of nitroarenes
Pandarus V, et al.
Advanced Synthesis & Catalysis, 353(8), 1306-1316 (2011)
A new class of heterogeneous platinum catalysts for the chemoselective hydrogenation of nitroarenes
Pandarus V, et al.
advanced synthesis and catalysis, 353(8), 1306-1316 (2011)
Yueer Yan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(34), 12161-12170 (2015-07-16)
A clear and deep understanding of zeolite crystallization with the addition of organosilane is desirable for the reasonable design and preparation of hierarchical zeolites. Herein, the effects of different organosilanes on zeolite crystallization were systematically studied. It was found that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service