Skip to Content
Merck
All Photos(1)

Key Documents

64258

Sigma-Aldrich

Dimethyl methylphosphonate

purum, ≥97.0% (GC)

Synonym(s):

Dimethoxymethyl phosphine oxide, Fyrol DMMP, Methanephosphonic acid dimethyl ester, NSC 62240, Reoflam DMMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3P(O)(OCH3)2
CAS Number:
Molecular Weight:
124.08
Beilstein:
878263
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.1 mmHg ( 20 °C)

Quality Level

grade

purum

Assay

≥97.0% (GC)

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.413 (lit.)
n20/D 1.413

bp

181 °C (lit.)

density

1.145 g/mL at 25 °C (lit.)

functional group

phosphonate

SMILES string

COP(C)(=O)OC

InChI

1S/C3H9O3P/c1-5-7(3,4)6-2/h1-3H3

InChI key

VONWDASPFIQPDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethyl methylphosphonate (DMPP) is generally used as a model compound for catalytic degradation studies due to its structural similarity to the organophosphorus compounds used in chemical warfare agents, pesticides and herbicides.
Used to test SXFA-coated SAW chemical sensors for organophosphorous compound detection
Used as a chemical agent simulant for testing of protective performance of PSS membranes and functionalized sorbent membranes

Reactant for photocatalytic oxidation

Additive for synthesis of:
  • Flame retardant high phosphorous containing unsaturated polyester resin
  • UV-cured epoxy acrylate / microencapsulated phase change material

Other Notes

Convenient Wittig-Horner reagent

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced photocatalytic degradation of dimethyl methylphosphonate in the presence of low-frequency ultrasound
Chen Y-C, et al.
Photochemical & Photobiological Sciences : Official Journal of the European Photochemistry Association and the European Society for Photobiology, 2(6), 694-698 (2003)
M. Mikolajczyk et al.
The Journal of Organic Chemistry, 43, 473-473 (1978)
Study of the catalytic destruction of dimethyl methylphosphonate (DMMP): oxidation over molybdenum (110)
Smentkowski VS, et al.
The Journal of Physical Chemistry, 92(22), 6351-6357 (1988)
Cristina Tudisco et al.
Langmuir : the ACS journal of surfaces and colloids, 28(3), 1782-1789 (2011-12-22)
This paper reports on the preparation of a porous silicon-based material covalently functionalized with cavitand receptors suited for the detection of organophosphorus vapors. Two different isomeric cavitands, both containing one acid group at the upper rim, specifically designed for covalent
Liangming Wei et al.
Nanotechnology, 22(42), 425501-425501 (2011-09-22)
We developed a chemiresistive sensor based on doped and functionalized semiconducting single-walled carbon nanotube (SWNT) networks for ultrasensitive and rapid detection of dimethyl methylphosphonate (DMMP) (simulant of nerve agent sarin) vapor. The semiconducting SWNT network was deposited between interdigitated electrodes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service