Skip to Content
Merck
All Photos(4)

Key Documents

398160

Sigma-Aldrich

Hexamethylenetetramine

ACS reagent, ≥99.0%

Synonym(s):

1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, Hexamine, Methenamine, Urotropine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N4
CAS Number:
Molecular Weight:
140.19
Beilstein:
2018
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99.0%

form

solid

autoignition temp.

770 °F

ign. residue

≤0.1%

loss

≤2.0% loss on drying

mp

280 °C (subl.) (lit.)

solubility

water: soluble

cation traces

heavy metals: ≤0.001% (by ICP)

SMILES string

C1N2CN3CN1CN(C2)C3

InChI

1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2

InChI key

VKYKSIONXSXAKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexamethylenetetramine (HMTA) is a water soluble, heterocyclic organic compound with a cage-like structure resembling adamantane. It is commonly used in treating urinary tract infection.

Application

Hexamethylenetetramine (HMTA) may be used as:
  • Raw material in the synthesis of blue emitting boron carbon oxynitride (BCNO) phosphors and Zinc oxide (ZnO) nanorods.
  • Catalyst in the synthesis of 2-amino-4H-pyran derivatives.
  • Surfactant in the synthesis of HMTA capped transition metals doped ZnS nanoparticles.
  • Precipitant and template in the synthesis of Ni(OH)2.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Skin Sens. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

250 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controlling Diameter, Length and Characterization of ZnO Nanorods by Simple Hydrothermal Method for Solar Cells.
Kurda AH, et al.
World Journal of Nano Science and Engineering, 5(1), 34-34 (2015)
Preparation of NiO Microspheres by Hydrothermal Method and its Electrochemical Capacitive Properties.
Li QQ, et al.
Materials Science Forum, 814, 81-85 (2015)
Blue emitting BCNO phosphors with high quantum yields.
Zhang X, et al.
Journal of Material Chemistry C, 3(14), 3311-3317 (2015)
Optical and photoluminescence properties of HMTA capped transition metals (Cu, Co and Mn) doped ZnS nanoparticles.
Anand KV.
Journal of Chemical and Pharmaceutical Research, 7(3), 286-290 (2015)
Methenamine in dairy products by isotope dilution gas chromatography coupled with triple quadrupole mass spectrometry: Method validation and occurrence.
Xu X, et al.
Food Control, 57, 89-95 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service