Skip to Content
Merck
All Photos(1)

Key Documents

O8801

Sigma-Aldrich

Oxalyl chloride

reagent grade, 98%

Synonym(s):

Ethanedioyl dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOCOCl
CAS Number:
Molecular Weight:
126.93
Beilstein:
1361988
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Quality Level

vapor density

4.4 (vs air)

vapor pressure

150 mmHg ( 20 °C)

Assay

98%

reaction suitability

reagent type: oxidant

impurities

≤1.0% phosgene content

refractive index

n20/D 1.429 (lit.)

bp

62-65 °C (lit.)

mp

−10-−8 °C (lit.)

density

1.5 g/mL at 20 °C (lit.)

SMILES string

ClC(=O)C(Cl)=O

InChI

1S/C2Cl2O2/c3-1(5)2(4)6

InChI key

CTSLXHKWHWQRSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxalyl chloride is a versatile reagent in various chemical transformations, such as chlorination, dichlorination, oxidation, reduction, dehydration, decarboxylation, formylation, and ring cleavage of epoxides.

Application

Oxalyl chloride can be used as an oxidizing agent:
  • To synthesize β, β′-diketodithioethers from β, β′-dihydroxydithioethers via Swern oxidation.
  • In the DMSO-catalyzed Swern oxidation of primary amides or aldoximes to nitriles in the presence of triethylamine as a base.
  • In the Moffatt-Swern oxidation of aryl allylic alcohols to halogenated unsaturated ketones in the presence of triethylamine.
Reactant involved in:
  • Synthesis of N-heterocyclic ynones and ynediones, used to activate carboxylic acids
  • Chlorination and halogenation
  • Three-component [3+2] cycloadditions
  • Reactions with organostannanes
  • Synthesis of cyclopentenones
  • Carbonylations, used as a carbonyl synthon

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxalyl chloride as a practical carbon monoxide source for carbonylation reactions.
Hansen SVF and Ulven T.
Organic Letters, 17(11), 2832-2835 (2015)
Oxalyl halides: Part II. Vibrational spectra and assignments for oxalyl fluoride and oxalyl chloride fluoride.
Hencher JL and King GW.
Journal of Molecular Spectroscopy, 16(1), 168-178 (1965)
A direct access to α-diones from oxalyl chloride.
Babudri F, et al.
Tetrahedron Letters, 36(40), 7305-7308 (1995)
Systematic survey of positive chlorine sources in the asymmetric Appel reaction: oxalyl chloride as a new phosphine activator.
Rajendran KV, et al.
Tetrahedron Letters, 54(51), 7009-7012 (2013)
Taisuke Itaya et al.
Chemical & pharmaceutical bulletin, 50(1), 83-86 (2002-02-05)
The relationship between the product patterns and the configurations of 1,2-cycloheptane- and 1,2-cyclooctanediols 9 in the cyclocondensations with oxalyl chloride in the presence of triethylamine at 0 degrees C has been shown analogous to that obtained for 1,2-disubstituted acyclic ethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service