Skip to Content
Merck
All Photos(1)

Key Documents

36143

Supelco

Malathion

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19O6PS2
CAS Number:
Molecular Weight:
330.36
Beilstein:
1804525
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)CC(SP(=S)(OC)OC)C(=O)OCC

InChI

1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3

InChI key

JXSJBGJIGXNWCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Malathion is a commonly used organophosphate[1] pesticide[2], which is generally sprayed on aquatic habitats in order to have a control over mosquitoes that carry malaria and the west nile virus[3].

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidative stress and cholinesterase inhibition in saliva and plasma of rats following subchronic exposure to malathion
Abdollahi M, et al.
Comparative Biochemistry and Physiology. Toxicology & Pharmacology : CBP, 137, 29-34 (2004)
Synergistic impacts of malathion and predatory stress on six species of North American tadpoles
Relyea.AR
Environmental Toxicology and Chemistry / Setac, 23, 1080-1084 (2004)
Removal of lindane and malathion from wastewater using bagasse fly ash?a sugar industry waste
Gupta.KV, et al.
Water Research, 36, 2483-2490 (2002)
Maya L Groner et al.
Ecological applications : a publication of the Ecological Society of America, 23(6), 1443-1454 (2013-10-24)
For the past several decades, amphibian populations have been decreasing around the globe at an unprecedented rate. Batrachochytrium dendrobatidis (Bd), the fungal pathogen that causes chytridiomycosis in amphibians, is contributing to amphibian declines. Natural and anthropogenic environmental factors are hypothesized
Guang-Mao Shen et al.
Pest management science, 68(12), 1553-1563 (2012-09-26)
The oriental fruit fly, Bactrocera dorsalis, is a major pest that infects fruits and agricultural products worldwide. The latest resistance monitoring of B. dorsalis from mainland China has identified high levels of resistance to insecticides. In this study, the biochemical

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

  1. Can you please provide me with the density or the volume contained in the vial with the specified concentration?

    1 answer
    1. The reported density is 1.23 g/mL at 25°C. For the 100 mg size, the volume of malathion is approximately 81.3 uL. With a molecular weight of 330.36 g/mole, the concentration is calculated to be 3.72 M.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service