Skip to Content
Merck
All Photos(1)

Key Documents

51976

Sigma-Aldrich

Ethyl alcohol, pure

≥99.8%, for molecular biology, (absolute alcohol, without additive, A15 o1), BioUltra

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
EC Number:
MDL number:
UNSPSC Code:
12191601
PubChem Substance ID:
NACRES:
NA.25

product name

Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)

grade

for molecular biology

Quality Level

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

product line

BioUltra

Assay

≥99.8%

form

liquid

autoignition temp.

683 °F

quality

(absolute alcohol, without additive, A15 o1)

expl. lim.

19 %, 60 °F

impurities

DNases, none detected
KMnO4-reducing substances, in accordance
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
substances darkened by H2SO4, in accordance
≤0.0004% free alkali (as NH3)
≤0.001% aldehyde (as CH3CHO)
≤0.001% formaldehyde (as HCHO)
≤0.001% ketone (as CH3COCH3)
≤0.002% free acid (as CH3COOH)
≤0.1% methanol (CH3OH)
≤0.2% water

evapn. residue

≤0.001%

refractive index

n20/D 1.3600 (lit.)
n20/D 1.362

bp

78 °C (lit.)

mp

−114 °C (lit.)

solubility

H2O: miscible
organic solvents: miscible

density

0.789 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.1 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCO

λ

neat

UV absorption

λ: 260 nm Amax: 0.04
λ: 280 nm Amax: 0.03

suitability

suitable for molecular biology

application(s)

life science and biopharma

format

neat

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethanol, also known as Ethyl Alcohol, is a volatile primary alcohol with a straight-chain structure. It is frequently employed as a biochemical reagent, effectively denaturing proteins from bacteria, fungi, and viruses. Widely utilized in cell biology, molecular biology, and biochemical research, this DNase, RNase, NICKase, and protease-free form of ethanol plays a crucial role in purifying and precipitating biomolecules like nucleic acids (DNA & RNA) and proteins. Additionally, it finds application in histology for preparing staining and destaining reagents, as well as in the dehydration of tissues before embedding.

Application

Ethanol has been used:
  • in vehicle-treated controls to perform mouse experiments
  • in viability assay along with crystal violet to stain human and mouse high-grade serous ovarian cancer (HGSOC) cells after stimulation
  • to prepare 17β-estradiol (E2) to use in cell culture experiments
  • in cell cycle analysis for fixing cells drop-wise

Biochem/physiol Actions

Ethanol is a widely used organic chemical in industry and consumer goods. This aliphatic alcohol is mainly used in industry as a solvent and as an intermediate in the production of other chemicals.

Features and Benefits

  • Versatile and adaptable for a wide variety of laboratory and research applications
  • BioUltra Grade for your Molecular Biology, Cell Biology and Biochemical research
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • Free from DNase, RNase, NICKase and proteaseTested for trace levels of Anions and Cations

Other Notes

For additional information on our range of Biochemicals, please complete this form.
Sales restrictions may apply. Suitable for the precipitation of nucleic acids

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

also commonly purchased with this product

Product No.
Description
Pricing

comparable product

Product No.
Description
Pricing

greener alternative product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

M G Mahoney et al.
The Journal of clinical investigation, 103(4), 461-468 (1999-02-18)
Patients with pemphigus foliaceus (PF) have blisters on skin, but not mucous membranes, whereas patients with pemphigus vulgaris (PV) develop blisters on mucous membranes and/or skin. PF and PV blisters are due to loss of keratinocyte cell-cell adhesion in the
D M Lovinger et al.
Science (New York, N.Y.), 243(4899), 1721-1724 (1989-03-31)
The ion current induced by the glutamate receptor agonist N-methyl-D-aspartate (NMDA) in voltage-clamped hippocampal neurons was inhibited by ethanol (EtOH). Inhibition increased in a concentration-dependent manner over the range 5 to 50 mM, a range that also produces intoxication. The
G M Huang et al.
Analytical biochemistry, 223(1), 35-38 (1994-11-15)
Genome projects require a high-throughput method for DNA template preparation, which traditional protocols can rarely provide. We introduce here a new protocol for plasmid and cosmid DNA preparation based on alkaline denaturation in a 96-well format. The essential improvement made
Preparation and properties of serum and plasma proteins; crystallization of serum albumins from ethanol water mixtures.
E J COHN et al.
Journal of the American Chemical Society, 69(7), 1753-1761 (1947-07-01)
Y Gavrieli et al.
The Journal of cell biology, 119(3), 493-501 (1992-11-01)
Programmed cell death (PCD) plays a key role in developmental biology and in maintenance of the steady state in continuously renewing tissues. Currently, its existence is inferred mainly from gel electrophoresis of a pooled DNA extract as PCD was shown

Articles

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service