Skip to Content
Merck
All Photos(4)

Key Documents

1.11727

Supelco

Ethyl alcohol, pure

greener alternative

≥99.9% (GC), gradient grade, suitable for HPLC, gradient grade, LiChrosolv®

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-578-6
NACRES:
NA.03
technique(s):
HPLC: suitable
bp:
78 °C (lit.)
vapor pressure:
44.6 mmHg ( 20 °C)
59 hPa ( 20 °C)

Product Name

Ethanol, gradient grade for liquid chromatography LiChrosolv®

grade

gradient grade
gradient grade

Quality Level

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)
59 hPa ( 20 °C)

product line

LiChrosolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

683 °F

potency

6200 mg/kg LD50, oral (Rat)

expl. lim.

19 %, 60 °F

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.1% Water

evapn. residue

≤2.0 mg/L

transmittance

225 nm, ≥60%
240 nm, ≥85%
260 nm, ≥98%

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L in H2O)

bp

78 °C (lit.)

mp

−114 °C (lit.)

transition temp

flash point 12 °C

format

neat

greener alternative category

storage temp.

2-30°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Ethanol is environmentally preferable green solvent and thus it aligns with "Safer Solvents and Auxiliaries". and "Use of Renewable feedstocks". Click here for more information.

With their high degree of UV transmittance, low particle count, low acidity and alkalinity, and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient” and "isocratic" grades. This can minimize the gradient effect of the solvent involved.

Application

Ethanol is used as a solvent:
  • to Prepare stock standard and working solutions of fenpropathrin and bifenthrin.
  • in alternative and sustainable solvents for green analytical chemistry.

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alternative and sustainable solvents for green analytical chemistry.
Ombeline C, et al.
Current opinion in green and sustainable chemistry., 31, 100510-100510 (2021)
Enantiomeric resolution of pyrethroids by high-performance liquid chromatography with diode-laser polarimetric detection.
Snchez, F Garca et al.
Journal of Chromatography A, 754, 97-102 (1996)

Articles

An RP-LC×RP-LC-PDA-MS method for the metabolic profiling of G. glabra extract using the MSG approach for enhanced metabolic coverage.

An RP-LC×RP-LC-PDA-MS method for the metabolic profiling of G. glabra extract using the MSG approach for enhanced metabolic coverage.

An RP-LC×RP-LC-PDA-MS method for the metabolic profiling of G. glabra extract using the MSG approach for enhanced metabolic coverage.

An RP-LC×RP-LC-PDA-MS method for the metabolic profiling of G. glabra extract using the MSG approach for enhanced metabolic coverage.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service