Skip to Content
Merck
All Photos(1)

Key Documents

02483

Supelco

Ethanol

standard for GC

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
standard for GC

Quality Level

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

Assay

≥99.9% (GC)

autoignition temp.

683 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

19 %, 60 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.3600 (lit.)
n20/D 1.362

bp

78 °C (lit.)

mp

−114 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethanol is a clear, colorless liquid, which can prepared by fermentation of certain carbohydrates. It is the primary ingredient in alcoholic beverages, and is widely used as a solvent and preservative in pharmaceutical preparations. As a solvent of substances intended for human contact or consumption, it is widely used in colorings, scents, flavorings, and medicines. Ethanol has a depressive effect on the central nervous system (CNS), and owing to its psychoactive effects, it is considered a drug. Ethanol has a complex mode of action and affects multiple systems in the brain, most notably it acts as an agonist to the GABA receptors. It is also a gasoline additive and mostly used as an alternative to petroleum as it can reduce CO and CO2 levels during emission when compared to other fossil fuels.

Application

Ethanol has been used as internal standard in GC-MS analysis for the determination of methanol and free glycerol in biodiesel. It may have been used as reference standard in GC analysis for determination of ethanol in beer.

Other Notes

Sales restrictions may apply

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simultaneous gas chromatographic determination of methanol and free glycerol in biodiesel
Mittelbach, M., Roth, G; and Bergmann, A.
Chromatographia, 42.7, 431-434 (1996)
Determination of ethanol in beer by direct injection gas chromatography: a comparison of six identical systems
Clarkson, S.P., Ormrod, I.H.L., Sharpe, F.R.
Journal of the Institute of Brewing, 101.3, 191-193 (1995)
S Enocksson et al.
The Journal of clinical investigation, 95(5), 2239-2245 (1995-05-01)
Although it is well established in several mammalian species that beta 3-adrenoceptors play a major role in regulating lipolysis and thermogenesis in adipose tissue, the functional existence and role of this receptor subtype in man has been controversial. We investigated
Gian Marco Leggio et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(8), 2017-2028 (2014-03-04)
Mesolimbic dopamine (DA) controls drug- and alcohol-seeking behavior, but the role of specific DA receptor subtypes is unclear. We tested the hypothesis that D3R gene deletion or the D3R pharmacological blockade inhibits ethanol preference in mice. D3R-deficient mice (D3R(-/-)) and
N Di Girolamo et al.
Stem cells (Dayton, Ohio), 33(1), 157-169 (2014-06-27)
Stem cell (SC) division, deployment, and differentiation are processes that contribute to corneal epithelial renewal. Until now studying the destiny of these cells in a living mammal has not been possible. However, the advent of inducible multicolor genetic tagging and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service