Skip to Content
Merck
All Photos(1)

Key Documents

36158

Supelco

Methidathion

PESTANAL®, analytical standard

Synonym(s):

S-2,3-Dihydro-5-methoxy-2-oxo-1,3,4-thiodiazol-3-yl-methyl-O,O-dimethyl-phosphodithioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N2O4PS3
CAS Number:
Molecular Weight:
302.33
Beilstein:
619915
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COC1=NN(CSP(=S)(OC)OC)C(=O)S1

InChI

1S/C6H11N2O4PS3/c1-10-5-7-8(6(9)16-5)4-15-13(14,11-2)12-3/h4H2,1-3H3

InChI key

MEBQXILRKZHVCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methidathion is a dimethoxyorganic phosphorus pesticide and cholinesterase inhibitor.

Application

Methidathion may be used as a reference standard for the determination of methidathion in serum and urine samples of acute poisoning by high-performance liquid chromatography with diode-array detector.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of the carcinogenic potential of pesticides: 2. Methidathion.
Quest J A, et al.
Regulatory Toxicology and Pharmacology, 12(2), 117-126 (1990)
Determination of organophosphorus pesticides in biological samples of acute poisoning by HPLC with diode-array detector.
Cho Y, et al.
Chemical & Pharmaceutical Bulletin, 45(4), 737-740 (1997)
Idriss Bakas et al.
Analytica chimica acta, 734, 99-105 (2012-06-19)
A specific adsorbent for extraction of methidathion from olive oil was developed. The design of the molecularly imprinted polymer (MIP) was based on the results of the computational screening of the library of polymerisable functional monomers. MIP was prepared by
Turhan Yavuz et al.
Toxicology letters, 155(1), 59-64 (2004-12-09)
We have examined the effect of subchronic methidathion (MD) administration on vascular wall damage. The experimental groups were: control group and rats treated with 5 mg/kg MD. The MD group was given MD by gavage for 5 days a week
R Sutcu et al.
Cell biology and toxicology, 22(3), 221-227 (2006-04-04)
Methidathion (MD) phosphorodithioic acid S-[(5-methoxy-2-oxo-1,3,4-thiadiazol-3(2H)-yl)methyl] O,O-dimethyl ester is the organophosphate insecticide (OPI) most commonly used worldwide in the pest control of crops. Subchronic MD exposure was evaluated for its effects on lipid peroxidation, the serum activities of cholinesterase (ChE), and

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service