Skip to Content
Merck
All Photos(1)

Key Documents

46027

Supelco

Dimethomorph

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22ClNO4
CAS Number:
Molecular Weight:
387.86
Beilstein:
8794474
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of E + Z isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1ccc(cc1OC)\C(=C\C(=O)N2CCOCC2)c3ccc(Cl)cc3

InChI

1S/C21H22ClNO4/c1-25-19-8-5-16(13-20(19)26-2)18(15-3-6-17(22)7-4-15)14-21(24)23-9-11-27-12-10-23/h3-8,13-14H,9-12H2,1-2H3/b18-14+

InChI key

QNBTYORWCCMPQP-NBVRZTHBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethomorph, a cinnamic acid derivative, is a systemic morpholine fungicide mainly used against peronosporomycetes plant pathogens.

Application

Dimethomorph may be used as a reference standard for the determination of dimethomorph in dried hops by solvent extraction and liquid chromatography with ultraviolet detection (LC-UV).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D H Young et al.
Pest management science, 57(11), 1081-1087 (2001-11-28)
Laboratory studies were conducted to evaluate the risk of developing field resistance to zoxamide, a new Oomycete fungicide which acts on microtubules. Zoxamide, metalaxyl and dimethomorph were compared with respect to the ease with which fungicide-resistant mutants could be isolated
Rachel Dosnon-Olette et al.
The Science of the total environment, 408(10), 2254-2259 (2010-02-17)
Aquatic plants take up, transform and sequester organic contaminants and may therefore be used in phytoremediation for the removal of pollutants from wastewaters. A better understanding of factors affecting the rate of contaminant uptake by aquatic plants is needed to
Phototransformation pathways of the fungicide dimethomorph ((E, Z) 4-[3-(4-chlorophenyl)-3-(3, 4-dimethoxyphenyl)-1-oxo-2-propenyl] morpholine), relevant to sunlit surface waters.
Avetta P, et al.
The Science of the Total Environment, 500, 351-360 (2014)
Field optimization of dimethomorph for the control of potato late blight Phytophthora infestans: application rate, interval, and mixtures.
Stein J M and Kirk W W
Crop Protection, 22(4), 609-614 (2003)
Biological mode of action of dimethomorph on Pseudoperonospora cubensis and its systemic activity in cucumber.
Wang H-C, et al.
Agricultural sciences in China, 8(2), 172-181 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service