Skip to Content
Merck
All Photos(1)

Key Documents

45688

Supelco

Thiophanate-methyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O4S2
CAS Number:
Molecular Weight:
342.39
Beilstein:
937942
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COC(=O)NC(=S)Nc1ccccc1NC(=S)NC(=O)OC

InChI

1S/C12H14N4O4S2/c1-19-11(17)15-9(21)13-7-5-3-4-6-8(7)14-10(22)16-12(18)20-2/h3-6H,1-2H3,(H2,13,15,17,21)(H2,14,16,18,22)

InChI key

QGHREAKMXXNCOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiophanate-methyl is a systemic benzimidazole fungicide, commonly used in agriculture for the pre- and post-harvest treatment of pathogen affecting a variety of fruits and vegetables.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thiophanate-methyl may be used as an analytical reference standard for the quantification of the analyte in fruits, and wines using different chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of carbendazim, thiabendazole and thiophanate-methyl in banana (Musa acuminata) samples imported to Italy
Veneziano A, et al.
Food Chemistry, 87(3), 383-386 (2004)
Darshan M Rudakiya et al.
International journal of biological macromolecules, 144, 586-595 (2019-12-13)
Laccase from previously reported hardwood degrading fungus, Tricholoma giganteum AGDR1, was isolated, identified at molecular level, biochemically characterized and also utilized for pesticide degradation. Laccase gene is comprised of 3752 bp, which encompassed 742-bp of 5' flanking upstream sequence with 12
Enoch A Osekre et al.
Journal of economic entomology, 102(3), 887-896 (2009-07-21)
Cotton hardlock caused by Fusarium verticillioides (Sacc. Nirenberg) can reduce cotton, Gossypium hirsutum L., yields > 70% in the southeastern United States. The spores infect flowers on the day of pollination, resulting in hardlock, which is the failure of the
Mariusz Cycoń et al.
Biodegradation, 22(3), 573-583 (2010-10-27)
Degradation of the fungicide thiophanate-methyl (TM) by Enterobacter sp. TDS-1 and Bacillus sp. TDS-2 isolated from sandy soil previously treated with TM was studied in mineral salt medium (MSM) and soil. Both strains were able to grow in MSM supplemented
Sufang Fan et al.
Bulletin of environmental contamination and toxicology, 90(4), 477-481 (2012-12-18)
To select representative crop among leaf vegetables which may contain the highest residue after fungicide uses, open field applications with thiophanate-methyl on six crops including pakchoi, rape, crown daisy, amaranth, spinach and lettuce were designed and conducted. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service