Skip to Content
Merck
All Photos(1)

Key Documents

34016

Supelco

Ketoprofen

VETRANAL®, analytical standard

Synonym(s):

2-(3-Benzoylphenyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O3
CAS Number:
Molecular Weight:
254.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C(O)=O)c1cccc(c1)C(=O)c2ccccc2

InChI

1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)

InChI key

DKYWVDODHFEZIM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Ketoprofen is a non-steroidal anti-inflammatory drug.

Application

Ketoprofen may be used as a reference standard in the determination of ketoprofen in plasma samples using reversed-phase high-performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Non-steroidal anti-inflammatory compound that is selective for COX-1.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of microspheres containing the anti-inflammatory agents, indomethacin, ibuprofen, and ketoprofen
Bodmeier R and Chen H
Journal of Controlled Release : Official Journal of the Controlled Release Society, 10(2), 167-175 (1989)
Zeting Yuan et al.
International journal of pharmaceutics, 446(1-2), 191-198 (2013-02-21)
Chitosan (CS) grafted with β-cyclodextrin (CD-g-CS) nanoparticles as a new carrier for poorly water-soluble drugs has been developed. The CD-g-CS polymer is readily synthesized from chitosan and mono-6-deoxy-6-(p-toluenesulfonyl)-β-cyclodextrin. Three different degrees of substitution (DS) of β-cyclodextrin (β-CD) on CD-g-CS (9.6
Yan-Ping Duan et al.
The Science of the total environment, 447, 267-273 (2013-02-09)
The occurrence and multi-phase distribution of five pharmaceutical compounds were investigated in an urban wastewater treatment plant (WWTP) receiving river by analysis of pharmaceuticals in sediment, particulate matter, conventional dissolved phase (>0.7 μm), colloidal phase (5 kDa to 0.7 μm)
Ikram Ullah Khan et al.
International journal of pharmaceutics, 441(1-2), 809-817 (2012-12-26)
Microchannels based microfluidic systems are able to obtain monodispersed microparticles but are limited by cost, time and channel clogging. We succeeded in on the fly encapsulation of high ketoprofen contents in acrylate-based copolymer microbeads by environment friendly UV induced free
Nathalie C Newby et al.
Journal of dairy science, 96(3), 1511-1520 (2013-01-22)
Surgical correction of left displaced abomasum (LDA) is common in lactating dairy cattle. Despite the growing acceptance that abdominal surgery is painful, few cows are administered analgesia following LDA surgery. The objective of this research was to examine the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service