
Ketoprofen

Synonym(s):
2-(3-Benzoylphenyl)propionic acid, Ketoprofen
Empirical Formula (Hill Notation):
C16H14O3
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing