Skip to Content
Merck
All Photos(2)

Key Documents

471909

Sigma-Aldrich

(S)-(+)-Ketoprofen

99%

Synonym(s):

(S)-(+)-3-Benzoyl-α-methylbenzeneacetic acid, (S)-2-(3-Benzoylphenyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COC6H4CH(CH3)CO2H
CAS Number:
Molecular Weight:
254.28
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]22/D +49°, c = 1 in methanol

mp

75-78 °C (lit.)

SMILES string

C[C@H](C(O)=O)c1cccc(c1)C(=O)c2ccccc2

InChI

1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)/t11-/m0/s1

InChI key

DKYWVDODHFEZIM-NSHDSACASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neeraj Mahindroo et al.
Journal of medicinal chemistry, 52(14), 4277-4287 (2009-06-24)
We report novel inhibitors of Gli1-mediated transcription as potential anticancer agents. Focused chemical libraries were designed and assessed for inhibition of functional cell-based Gli1-mediated transcription and selective toxicity toward cancer cells. The SAR was revealed, and the selectivity of the
Austin D Hocker et al.
eLife, 8 (2019-03-23)
Neonatal inflammation is common and has lasting consequences for adult health. We investigated the lasting effects of a single bout of neonatal inflammation on adult respiratory control in the form of respiratory motor plasticity induced by acute intermittent hypoxia, which
Kathrin Renner et al.
Cell reports, 29(1), 135-150 (2019-10-03)
Tumor-derived lactic acid inhibits T and natural killer (NK) cell function and, thereby, tumor immunosurveillance. Here, we report that melanoma patients with high expression of glycolysis-related genes show a worse progression free survival upon anti-PD1 treatment. The non-steroidal anti-inflammatory drug
Marcello Allegretti et al.
Journal of medicinal chemistry, 48(13), 4312-4331 (2005-06-25)
The CXC chemokine CXCL8/IL-8 plays a major role in the activation and recruitment of polymorphonuclear (PMN) cells at inflammatory sites. CXCL8 activates PMNs by binding the seven-transmembrane (7-TM) G-protein-coupled receptors CXC chemokine receptor 1 (CXCR1) and CXC chemokine receptor 2
Juan M Padró et al.
Journal of separation science, 38(14), 2423-2430 (2015-05-02)
Valuable quantitative information could be obtained from strongly overlapped chromatographic profiles of two enantiomers by using proper chemometric methods. Complete separation profiles where the peaks are fully resolved are difficult to achieve in chiral separation methods, and this becomes a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service