Skip to Content
Merck
All Photos(2)

Key Documents

31712

Supelco

Fluazifop-P-butyl

PESTANAL®, analytical standard

Synonym(s):

Butyl (R)-2-[4-(5-trifluoromethyl-2-pyridyloxy)phenoxy]propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20F3NO4
CAS Number:
Molecular Weight:
383.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCOC(=O)[C@@H](C)Oc1ccc(Oc2ccc(cn2)C(F)(F)F)cc1

InChI

1S/C19H20F3NO4/c1-3-4-11-25-18(24)13(2)26-15-6-8-16(9-7-15)27-17-10-5-14(12-23-17)19(20,21)22/h5-10,12-13H,3-4,11H2,1-2H3/t13-/m1/s1

InChI key

VAIZTNZGPYBOGF-CYBMUJFWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluazifop-P-butyl belongs to the arylophenoxypropionate group of graminicides. It is a post-emergence herbicide that acts by inhibiting acetyl-CoA carboxylase enzyme activity, involved in the catalysis of the formation of malonyl-CoA during metabolism of lipids.

Application

Fluazifop-P-butyl may be used as a reference standard for the determination of the analyte:
  • In different varieties of lettuce (Lactuca sativum) by capillary gas chromatography with nitrogen–phosphorus detection (NPD).
  • In tea samples by modified QuEChERS (quick, easy, cheap, effective, rugged, and safe) sample preparation procedure combined with ultra-performance liquid chromatography-electrospray tandem mass spectrometry (UPLC/MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup - (External MSDS)

Flash Point(C)

50 °C - closed cup - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effect of fluazifop-p-butyl treatment on pigments and polyamines level within tissues of non-target maize plants.
Horbowicz M, et al.
Pesticide Biochemistry and Physiology, 107(1), 78- 85 (2013)
A multi-residue method for fast determination of pesticides in tea by ultra performance liquid chromatography- electrospray tandem mass spectrometry combined with modified QuEChERS sample preparation procedure.
Chen G, et al.
Food Chemistry, 125(4), 1406- 1411 (2011)
Simplified multiresidue method for determination of pesticide residues in lettuce by gas chromatography with nitrogen- phosphorus detection.
Fenoll J, et al.
Analytical and Bioanalytical Chemistry, 389(2), 643- 651 (2007)
Fluazifop-P-butyl (herbicide) affects richness and structure of soil bacterial communities.
Darine T, et al.
Soil Biology and Biochemistry, 81, 89- 97 (2015)
N W McCracken et al.
Biochemical pharmacology, 45(1), 31-36 (1993-01-07)
In the present study, the distribution and nature of esterases in the rat which hydrolysed fluazifop-butyl, carbaryl, paraoxon and phenylacetate were investigated. Vmax and Km values for the hydrolysis reactions were determined. Fluazifop-butyl was hydrolysed to fluazifop by rat liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service