Skip to Content
Merck
All Photos(2)

Key Documents

32824

Supelco

Fluazifop

PESTANAL®, analytical standard

Synonym(s):

2-[4-(5-Trifluoromethyl-2-pyridyloxy)phenoxy]propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12F3NO4
CAS Number:
Molecular Weight:
327.26
Beilstein:
552236
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(Oc1ccc(Oc2ccc(cn2)C(F)(F)F)cc1)C(O)=O

InChI

1S/C15H12F3NO4/c1-9(14(20)21)22-11-3-5-12(6-4-11)23-13-7-2-10(8-19-13)15(16,17)18/h2-9H,1H3,(H,20,21)

InChI key

YUVKUEAFAVKILW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Fluazifop is one of the major metabolites of fluazifop-p-butyl, a systemic, post-emergence herbicide used for the control of annual and perennial grass weeds in non-graminaceous crops.

Application

Fluazifop may be used as an analytical reference standard for the determination of the analyte in:
  • Rainwater samples by solid-phase extraction (SPE) and liquid chromatography-tandem mass spectrometry (LC-MS/MS).
  • Vegetables by acetonitrile-based QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction followed by LC-MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Persistence studies with the herbicide fluazifop-butyl in Saskatchewan soils under laboratory and field conditions.
A E Smith
Bulletin of environmental contamination and toxicology, 39(1), 150-155 (1987-07-01)
T Varga et al.
Acta veterinaria Hungarica, 47(1), 123-128 (1999-04-24)
The purpose of this work was to determine the individual and combined effects of insecticide Sumithion 50 EC (50% fenitrothion) and herbicide Fusilade S (12.5% fluazifop-P-butyl) on the development of pheasant embryos. Eggs were treated by injection of various concentrations
Residues of fluazifop-p-butyl following application to soybean.
G Kulshrestha et al.
Bulletin of environmental contamination and toxicology, 55(2), 276-282 (1995-08-01)
K A Walker et al.
The Biochemical journal, 254(1), 307-310 (1988-08-15)
Fluazifop is a grass-selective herbicide that appears to act by inhibiting fatty acid synthesis de novo in sensitive species. Results from four different types of experiment show that this inhibition is due to an action of fluazifop on acetyl-CoA carboxylase
Ahmed A Tantawy
Journal of the Egyptian Society of Parasitology, 32(3), 837-847 (2003-01-07)
The herbicides, Butachlor and Fluazifop-p-butyl were evaluated against B. alexandrina and their infection with S. mansoni, as well as against the miracidia and cercariae. The tested herbicides reduced the survival and infection rates of B. alexandrina by 28% and 35.71%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service