Skip to Content
Merck
All Photos(2)

Key Documents

45553

Supelco

Mancozeb

PESTANAL®, analytical standard

Synonym(s):

Manganese Zinc ethylenebis(dithiocarbamate)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

technical mixture

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

S=C1NCCNC(=S)S[Mn]S1.S=C2NCCNC(=S)S[Zn]S2

InChI

1S/2C4H8N2S4.Mn.Zn/c2*7-3(8)5-1-2-6-4(9)10;;/h2*1-2H2,(H2,5,7,8)(H2,6,9,10);;/q;;2*+2/p-4

InChI key

CHNQZRKUZPNOOH-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1B - Skin Sens. 1 - STOT RE 2

Target Organs

Nervous system,Endocrine system,Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Axelstad et al.
Toxicological sciences : an official journal of the Society of Toxicology, 120(2), 439-446 (2011-01-27)
The widely used fungicide mancozeb has been shown to cause hypothyroxinemia and other adverse effects on the thyroid hormone system in adult experimental animals. In humans, hypothyroxinemia early in pregnancy is associated with adverse effects on the developing nervous system
Shilpa Tyagi et al.
Omics : a journal of integrative biology, 15(3), 155-167 (2011-03-08)
Mancozeb, ethylene(bis)dithiocarbamate fungicides, has been well documented in the literature as a multipotent carcinogen, but the underlying mechanism remains unrevealed. Thus, mancozeb has been selected in this study with the objective to decipher the molecular mechanism that culminates in carcinogenesis.
Rekek Negga et al.
Neurotoxicology, 32(3), 331-341 (2011-03-08)
Epidemiological evidence suggests positive correlations between pesticide usage and the incidence of Parkinson's disease (PD). To further explore this relationship, we used wild type (N2) Caenorhabditis elegans (C. elegans) to test the following hypothesis: Exposure to a glyphosate-containing herbicide (TD)
Ana Gonzálvez et al.
Analytica chimica acta, 688(2), 191-196 (2011-02-22)
A methodology based on the vapour phase (VP) generation of carbon disulphide from non-volatile dithiocarbamate compounds has been developed for determination of pesticide residues in foodstuff at low ppm levels. The method involves a selective reaction combined with liquid phase
M Srinivasulu et al.
Environmental geochemistry and health, 34(3), 365-374 (2011-07-21)
The effect of selected pesticides, monocrotophos, chlorpyrifos alone and in combination with mancozeb and carbendazim, respectively, was tested on nitrification and phosphatase activity in two groundnut (Arachis hypogeae L.) soils. The oxidation of ammonical nitrogen was significantly enhanced under the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service