Skip to Content
Merck
All Photos(1)

Key Documents

36191

Supelco

Pendimethalin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19N3O4
CAS Number:
Molecular Weight:
281.31
Beilstein:
2157711
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCC(CC)Nc1c(cc(C)c(C)c1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C13H19N3O4/c1-5-10(6-2)14-12-11(15(17)18)7-8(3)9(4)13(12)16(19)20/h7,10,14H,5-6H2,1-4H3

InChI key

CHIFOSRWCNZCFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Morgane Danion et al.
Ecotoxicology and environmental safety, 79, 28-34 (2012-03-01)
In this study, the in vivo effects of chronic pollution by the active substance (AS) pendimethalin, a dinitroaniline herbicide, on the susceptibility of rainbow trout, Oncorhynchus mykiss L., to an experimental challenge with viral hemorrhagic septicemia virus (VHSV) were assessed.
Indu Chopra et al.
Environmental monitoring and assessment, 160(1-4), 123-126 (2009-01-09)
The mobility of pendimethalin in sandy loam soil was studied in soil columns under laboratory conditions at two application rates, 1.0 and 2.0 kg a.i. ha(-1), with simulated rainfall of 300 mm. The maximum concentration of the herbicide was found
Amal C Das et al.
Environmental monitoring and assessment, 184(4), 1985-1989 (2011-06-16)
An experiment was conducted under laboratory conditions to investigate the effect of two systemic herbicides viz., pendimethalin and quizalofop, at their recommended field rates (1.0 kg and 50 g active ingredient ha(- 1), respectively) on the growth and activities of non-symbiotic
Morgane Danion et al.
The Science of the total environment, 424, 143-152 (2012-03-27)
The in vivo effects of the herbicide active substance (AS) pendimethalin (alone and with Prowl 400® adjuvant) were evaluated on sanitary status i.e. the health status with regard to chemical pollution and on the physiological state via the immune system
U Undeğer et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(2), 502-508 (2009-11-10)
The endocrine disrupting potential of the herbicide pendimethalin was investigated in vivo on the uterotrophic response and on the expression of estrogen-regulated genes examined by quantitative real-time RT PCR. Receptor binding characteristics of pendimethalin were analyzed by an in silico

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service