Skip to Content
Merck
All Photos(1)

Key Documents

18909

Sigma-Aldrich

Calcofluor White Stain

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
MA.02

product line

BioChemika

Quality Level

form

liquid

composition

Calcofluor White M2R, 1 g/L
Evans blue, 0.5 g/L

technique(s)

microbe id | staining: suitable

color

blue to very dark blue

λmax

423-443 nm

fluorescence

λex 423-443 nm in 0.1 M phosphate pH 7.0

application(s)

food and beverages
hematology
histology

microbiology

storage temp.

room temp

suitability

fungi
parasites
yeasts

SMILES string

C1=CC=C(C=C1)NC2=NC(=NC(=N2)N(CCO)CCO)NC3=CC(=C(C=C3)C=CC4=C(C=C(C=C4)NC5=NC(=NC(=N5)NC6=CC=CC=C6)N(CCO)CCO)S(=O)(=O)O)S(=O)(=O)O

InChI

InChI=1S/C40H44N12O10S2/c53-21-17-51(18-22-54)39-47-35(41-29-7-3-1-4-8-29)45-37(49-39)43-31-15-13-27(33(25-31)63(57,58)59)11-12-28-14-16-32(26-34(28)64(60,61)62)44-38-46-36(42-30-9-5-2-6-10-30)48-40(50-38)52(19-23-55)20-24-56/h1-16,25-26,53-56H,17-24H2,(H,57,58,59)(H,60,61,62)(H2,41,43,45,47,49)(H2,42,44,46,48,50)/b12-11+

InChI key

CNGYZEMWVAWWOB-VAWYXSNFSA-N

General description

Calcofluor White Stain (C.l. 40622) is a stilbene compound that has additional triazinyl and phenyl aromatic groups. It is also known as cellufluor.

Application

Calcofluor White Stain is a fluorescent stain that is used for the rapid detection of yeasts, fungi, and parasitic organisms. It is a non-specific fluorochrome that binds to cellulose and chitin in cell walls. It is suitable for use as a viability stain for plant cells. Additionally, calcofluor white is recommended as a counterstain for in situ hybridization staining of plant material and as a flow cytometry stain for assessing the viability of animal cells.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas R Thomson et al.
Genome research, 18(10), 1624-1637 (2008-06-28)
We have determined the complete genome sequences of a host-promiscuous Salmonella enterica serovar Enteritidis PT4 isolate P125109 and a chicken-restricted Salmonella enterica serovar Gallinarum isolate 287/91. Genome comparisons between these and other Salmonella isolates indicate that S. Gallinarum 287/91 is
Bojan Gujas et al.
Development (Cambridge, England), 144(19), 3578-3589 (2017-08-31)
The plant vascular network consists of specialized phloem and xylem elements that undergo two distinct morphogenetic developmental programs to become transport-functional units. Whereas vacuolar rupture is a determinant step in protoxylem differentiation, protophloem elements never form a big central vacuole.
Hokyoung Son et al.
PloS one, 8(9), e72915-e72915 (2013-09-17)
Fusarium graminearum (teleomorph Gibberella zeae) is a prominent pathogen that infects major cereal crops such as wheat, barley, and maize. Both sexual (ascospores) and asexual (conidia) spores are produced in F. graminearum. Since conidia are responsible for secondary infection in
Aimee K Zaas et al.
PLoS genetics, 4(6), e1000101-e1000101 (2008-06-21)
Invasive aspergillosis (IA) is a common and life-threatening infection in immunocompromised individuals. A number of environmental and epidemiologic risk factors for developing IA have been identified. However, genetic factors that affect risk for developing IA have not been clearly identified.
Yifan Wu et al.
Nature communications, 10(1), 58-58 (2019-01-06)
Bloodborne infections with Candida albicans are an increasingly recognized complication of modern medicine. Here, we present a mouse model of low-grade candidemia to determine the effect of disseminated infection on cerebral function and relevant immune determinants. We show that intravenous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service