Skip to Content
Merck
All Photos(1)

Key Documents

C6767

Sigma-Aldrich

Congo Red

Dye content ≥35 %, Powder

Synonym(s):

Congo Red 4B, Cosmos Red, Cotton Red B, Cotton Red C, Direct Red 28, Direct Red R, Direct Red Y

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H22N6Na2O6S2
CAS Number:
Molecular Weight:
696.66
Colour Index Number:
22120
Beilstein:
3894858
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Congo Red, Dye content ≥35 %

form

powder

Quality Level

composition

Dye content, ≥35%

technique(s)

titration: suitable

color

red-brown

pH

6.7 (10 g/L)

mp

>360 °C (lit.)

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Nc1c(cc(c2ccccc12)S([O-])(=O)=O)\N=N\c3ccc(cc3)-c4ccc(cc4)\N=N\c5cc(c6ccccc6c5N)S([O-])(=O)=O

InChI

1S/C32H24N6O6S2.2Na/c33-31-25-7-3-1-5-23(25)29(45(39,40)41)17-27(31)37-35-21-13-9-19(10-14-21)20-11-15-22(16-12-20)36-38-28-18-30(46(42,43)44)24-6-2-4-8-26(24)32(28)34;;/h1-18H,33-34H2,(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2/b37-35+,38-36+;;

InChI key

IQFVPQOLBLOTPF-HKXUKFGYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Congo red has been used as a histochemical stain for the quantification of amyloid deposits in brain tissues.
  • It has been used as a cell wall inhibiting agent in fungus Cryptococcus neoformans.
  • It has also been used to study microstructural details in the cell walls of raw potato and fried potato using confocal microscopy.

Biochem/physiol Actions

Congo red is a benzidine-based anionic diazo dye. It is a histological dye which binds to many amyloid proteins and is used for the quantification of amyloid β-peptide aggregation. Congo red also interacts with β-D-glucans, polysaccharides containing continuous β-(1→4)-linked D-glucopyranosyl units and some hemicellulosic galactoglucomannans.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Some changes in potato chips during frying observed by confocal laser scanning microscopy (CLSM).
Pedreschi F and Aguilera JM
Food Science and Technology International = Ciencia y tecnologia de Los Alimentos Internacional, 8, 197-201 (2002)
Quantification of cerebral amyloid angiopathy and parenchymal amyloid plaques with Congo red histochemical stain.
Wilcock DM, et al.
Nature Protocols, 1, 1591-1591 (2006)
Kinetic study and equilibrium isotherm analysis of Congo Red adsorption by clay materials.
Chemical Engineering Journal, 148, 354-364 (2009)
Anat Frydman-Marom et al.
PloS one, 6(1), e16564-e16564 (2011-02-10)
An increasing body of evidence indicates that accumulation of soluble oligomeric assemblies of β-amyloid polypeptide (Aβ) play a key role in Alzheimer's disease (AD) pathology. Specifically, 56 kDa oligomeric species were shown to be correlated with impaired cognitive function in
PKC1 is essential for protection against both oxidative and nitrosative stresses, cell integrity, and normal manifestation of virulence factors in the pathogenic fungus Cryptococcus neoformans.
Gerik KJ, et al.
Eukaryotic Cell, 7, 1685-1685 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service