Skip to Content
Merck
All Photos(1)

Key Documents

10236276001

Roche

DAPI

4′,6-Diamidine-2′-phenylindole dihydrochloride

Synonym(s):

4′,6-Diamidino-2-phenylindole dihydrochloride, 2-(4-Amidinophenyl)-6-indolecarbamidine dihydrochloride, DAPI dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N5 · 2HCl
CAS Number:
Molecular Weight:
350.25
Beilstein:
4894417
MDL number:
UNSPSC Code:
41116100
PubChem Substance ID:

Quality Level

Assay

>90%

form

powder

packaging

pkg of 10 mg

manufacturer/tradename

Roche

λmax

340 nm in aq. suspension

fluorescence

λex 340 nm; λem 488 nm (nur DAPI)
λex 364 nm; λem 454 nm (DAPI-DNA-Komplex)

storage temp.

room temp

SMILES string

Cl.Cl.NC(=N)c1ccc(cc1)-c2cc3ccc(cc3[nH]2)C(N)=N

InChI

1S/C16H15N5.2ClH/c17-15(18)10-3-1-9(2-4-10)13-7-11-5-6-12(16(19)20)8-14(11)21-13;;/h1-8,21H,(H3,17,18)(H3,19,20);2*1H

InChI key

FPNZBYLXNYPRLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

DAPI is a fluorescent dye that binds selectively to double-stranded DNA and forms strongly fluorescent DNA-DAPI complexes with high specificity. It is commonly used to detect mycoplasma in cell culture via fluorescence microscopy.
DAPI is several times more sensitive than ethidium bromide for staining DNA in agarose gels. It may be used for photofootprinting of DNA, to detect annealed probes in blotting applications by specifically visualizing the double-stranded complex, and to study the changes in DNA and analyze DNA content during apoptosis using flow cytometry. DAPI staining has also been shown to be a sensitive and specific detection method for mycoplasma.

Biochem/physiol Actions

Cell permeable fluorescent minor groove-binding probe for DNA. Binds to the minor groove of double-stranded DNA (preferentially to AT rich DNA), forming a stable complex which fluoresces approximately 20 times greater than DAPI alone.

Quality

Purity: >90% (from N)

Principle

The fluorescent dye DAPI binds selectively to DNA and forms strongly fluorescent DNA-DAPI complexes with high specificity. DAPI, once added to tissue culture cells, is rapidly taken up into cellular DNA, yielding highly fluorescent nuclei and no detectable cytoplasmic fluorescence. When the cells are contaminated with Mycoplasmas, characteristic discrete fluorescent foci are readily detected over the cytoplasm and sometimes in intercellular spaces.

Preparation Note

Working solution: Solubility: 25 mg/ml in water


Preparation of stock solution


Dissolve in double-dist. water to a final concentration of 1 to 5 mg/ml.
Note: Do not use any buffers.


Preparation of working solution


Dilute the stock solution with methanol to a final concentration of 1 μg/ml. The working solution is stable at 2 to 8 °C for about 6 months.
Storage conditions (working solution): Stock solution (1 to 5 mg/ml) at -15 to -25 °C for 12 months.
Working solution (1μg/ml) at 2 to 8 °C for about 6 months.

Reconstitution

In 2 to 10 ml double-dist. water; 1 to 5 mg/ml final concentration.
Note: Prepare aliquots and store at -15 to -25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Itsuko Nihonmatsu et al.
Biology open, 9(1) (2019-12-26)
Late-phase long-term potentiation (L-LTP) in hippocampus, thought to be the cellular basis of long-term memory, requires new protein synthesis. Neural activity enhances local protein synthesis in dendrites, which in turn mediates long-lasting synaptic plasticity. Ca2+/calmodulin-dependent protein kinase IIα (CaMKIIα) is
Clara Dobler et al.
Cells, 9(9) (2020-09-05)
DNA damage response inhibitors (DDRi) may selectively enhance the inactivation of tumor cells in combination with ionizing radiation (IR). The induction of senescence may be the key mechanism of tumor cell inactivation in this combinatorial treatment. In the current study
Vahid Molla Kazemiha et al.
Cytotechnology, 68(4), 1063-1080 (2015-03-07)
Mycoplasmas are the most important contaminants of cell cultures throughout the world. They are considered as a major problem in biological studies and biopharmaceutical economic issues. In this study, our aim was to find the best standard technique as a
Xiaoyue Zhu et al.
Journal of neuroinflammation, 17(1), 78-78 (2020-03-05)
Cerebral amyloid angiopathy (CAA) is a common cerebral small vessel disease of the aged and a prominent comorbidity of Alzheimer's disease (AD). CAA can promote a variety of vascular-related pathologies including neuroinflammation, cerebral infarction, and hemorrhages, which can all contribute
Sabyasachi Sen et al.
Human gene therapy, 21(10), 1327-1334 (2010-05-26)
The regenerative potential of bone marrow-derived endothelial progenitor cells (EPCs) has been adapted for the treatment of myocardial and limb ischemia via ex vivo expansion. We sought to enhance EPC function by the efficient genetic modification of EPCs in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service