Skip to Content
Merck
All Photos(1)

Key Documents

83689

Sigma-Aldrich

Rhodamine B

for fluorescence

Synonym(s):

Basic Violet 10, Brilliant Pink B, Rhodamine O, Tetraethylrhodamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31ClN2O3
CAS Number:
Molecular Weight:
479.01
Colour Index Number:
45170
Beilstein:
4091619
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

grade

for fluorescence

Quality Level

form

powder or crystals

mp

210-211 (dec.) (lit.)

solubility

H2O: soluble
methanol: soluble

fluorescence

λex 553 nm; λem 627 nm in methanol

suitability

in accordance for fluorescence

SMILES string

[Cl-].CCN(CC)c1ccc2c(OC3=CC(\C=CC3=C2c4ccccc4C(O)=O)=[N+](/CC)CC)c1

InChI

1S/C28H30N2O3.ClH/c1-5-29(6-2)19-13-15-23-25(17-19)33-26-18-20(30(7-3)8-4)14-16-24(26)27(23)21-11-9-10-12-22(21)28(31)32;/h9-18H,5-8H2,1-4H3;1H

InChI key

PYWVYCXTNDRMGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rhodamine B is a xanthene dye, which functions as a water tracer fluorescent. It is used as a staining fluorescent dye.
Rhodamine B is bright-red in color.

Application

Rhodamine B has been used:
  • as a fluorescent dye for cross staining 1-N-methylamino-5-isocyanonaphthalene (MICAN)
  • as a fluorescent dye in confocal microscopy
  • to study its fluorescence in water and ethanol solutions

for cross staining of N-methylamino-5-isocyanonaphthalene (MICAN)

Analysis Note

λmax. ∼543 nm, E(1%/1cm) ∼2200-2800 (Methanol)

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The
A study of water circulation in Monterey harbor using rhodamine B dye (1965)
Simplifying microfluidic separation devices towards field-detection of blood parasites
Holm SH, et al.
Analytical Methods : Advancing Methods and Applications, 8(16), 3291-3300 (2016)
MICAN, a new fluorophore for vital and non-vital staining of human cells
Nagy Z, et al.
Toxicology in vitro, 48, 137-145 (2018)
The fluorescence properties of cationic rhodamine B in the gas phase
Sagoo SK and Jockusch RA
Journal of Photochemistry and Photobiology A: Chemistry, 220(2-3), 173-178 (2011)
Rhodamine B as a probe for phase transition in liquid solutions
Shishkin I, et al.
arXiv (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service