Skip to Content
Merck
All Photos(1)

Key Documents

02891

Sigma-Aldrich

Ethanol

purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.05

vapor density

1.59 (vs air)
1.59 (vs air)

Quality Level

vapor pressure

44.6 mmHg ( 20 °C)
44.6 mmHg

grade

purum

Assay

~96% (based on denaturant-free substance)

form

liquid

autoignition temp.

683 °F

quality

secunda spirit, denaturated with 2% 2-butanone, S15

expl. lim.

19 %, 60 °F
19 %

refractive index

n20/D 1.3600 (lit.)
n20/D 1.364

bp

78 °C (lit.)
78 °C

mp

-114 °C
−114 °C (lit.)

density

0.789 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
Sales restrictions may apply

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D L Jones et al.
European journal of pharmacology, 59(1-2), 11-21 (1979-10-26)
Oral motor responses were recorded in water-deprived rats receiving injections of spiroperidol, the dopamine blocker, into the nucleus accumbens and caudate nucleus. Spiroperidol injected into the nucleus accumbens resulted in dose dependent decreases in lap volume and tongue extension. Injections
Physicochemical Properties of SnO2-Sb2 O 5 Films Prepared by the Spray Pyrolysis Technique.
Correa-Lozano B, et al.
Journal of the Electrochemical Society, 143(1), 203-209 (1996)
50-Hz extremely low frequency electromagnetic fields enhance cell proliferation and DNA damage: possible involvement of a redox mechanism.
Wolf FI, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1743(1), 120-129 (2005)
Fabrication of thin film dye sensitized solar cells with solar to electric power conversion efficiency over 10%.
Ito S, et al.
Thin Solid Films, 516(14), 4613-4619 (2008)
Spectrophotometric characteristics of chlorophylls a and b and their pheophytins in ethanol.
J F Wintermans et al.
Biochimica et biophysica acta, 109(2), 448-453 (1965-11-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service