Skip to Content
Merck
All Photos(3)

Key Documents

MABE462

Sigma-Aldrich

Anti-TET2 Antibody, clone hT2H 21F11

clone hT2H21F11, from mouse

Synonym(s):

Methylcytosine dioxygenase TET2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

hT2H21F11, monoclonal

species reactivity

human, mouse

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TET2(54790)

General description

Methylcytosine dioxygenase TET2 catalyzes the conversion of methylcytosine (5mC) to 5-hydroxymethylcytosine (5hmC). The function of 5-hydroxymethylcytosine is currently unclear but it may influence chromatin structure, or act as an intermediate component in cytosine demethylation. TET2 is frequently mutated in myeloproliferative disorders (MPD) or myeloproliferative neoplasms (MPN), and systemic mastocytosis. TET2 disorders also cause polycythemia vera (PV) and myelodysplastic syndrome (MDS).

Specificity

This antibody recognizes the N-terminus of TET2

Immunogen

Recombinant protein corresponding to the N-terminus of human TET2.

Application

Detect Tet2 using this mouse monoclonal antibody, Anti-TET2 Antibody, clone hT2H 21F11 validated for use in Chromatin IP (ChIP), western blotting & IP.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected TET2 in 10 µg of TF-1 cell lysate.
Immunoprecipitation Analysis: 5 µg of a representative lot immunoprecipitated TET2 in 250 µg of HL60 RIPA cell lysate.

Quality

Evaluated by Western Blotting in MOLT-4 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected TET2 in 10 µg of MOLT-4 cell lysate.

Target description

~ 260 kDa observed. The calculated molecular weight of this protein is 224 kDa, but can be observed at ~260 kDa

Physical form

Format: Purified

Analysis Note

Control
MOLT-4 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zihang Zeng et al.
Redox biology, 67, 102916-102916 (2023-10-10)
Chronic obstructive pulmonary disease (COPD) is a significant global cause of morbidity and mortality currently. Long-term exposure of cigarette smoke (CS) inducing persistent inflammation, small airway remodeling and emphysematous lung are the distinguishing features of COPD. Ferroptosis, occurred in lung
Lei-Lei Chen et al.
Science advances, 6(38) (2020-09-20)
TET2 DNA dioxygenase is frequently mutated in human hematopoietic malignancies and functionally inactivated in many solid tumors through a nonmutational mechanism. We recently found that TET2 mediates the interferon-JAK-STAT pathway to stimulate chemokine expression and tumor infiltration of lymphocytes (TILs).
Yueyue Duan et al.
Journal of medical virology, 94(7), 3251-3256 (2022-02-26)
Swine acute diarrhea syndrome coronavirus (SADS-CoV) is a newly discovered bat-origin coronavirus with fatal pathogenicity for neonatal piglets. There is no vaccine to prevent SADS-CoV infection or clinically approved drugs targeting SADS-CoV. Therefore, unraveling cellular factors that regulate SADS-CoV for
Lijuan Luo et al.
iScience, 27(3), 109252-109252 (2024-03-05)
DNA demethylase TET2 was related with lung function. However, the precise role of TET2 in cigarette smoke (CS)-induced apoptosis of airway epithelium cells, and the mechanisms involved, have yet to be elucidated. Here, we showed that CS decreased TET2 protein
Qi Wang et al.
mBio, 10(4) (2019-08-23)
HIV-1 Vpr enhances viral replication in human macrophages via multiple mechanisms that are not clearly defined. It does not affect HIV-1 virion production during the first round of infection. We have recently discovered that Vpr targets the DNA demethylase TET2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service