Skip to Content
Merck
All Photos(3)

Key Documents

ABE364

Sigma-Aldrich

Anti-Tet2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Methylcytosine dioxygenase TET2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TET2(54790)

General description

Methylcytosine dioxygenase TET2 catalyzes the conversion of methylcytosine (5mC) to 5-hydroxymethylcytosine (5hmC). The function of 5-hydroxymethylcytosine is currently unclear but it may influence chromatin structure, or act as an intermediate component in cytosine demethylation. TET2 is frequently mutated in myeloproliferative disorders (MPD) or myeloproliferative neoplasms (MPN), and systemic mastocytosis. TET2 disorders also cause polycythemia vera (PV) and myelodysplastic syndrome (MDS).

Immunogen

KLH-conjugated linear peptide corresponding to human Tet2.

Application

Anti-Tet2 detects levels of Tet2 proteins & has been published & validated for use in WB, IHC & ICC.
Immunohistochemistry Analysis: A 1:4,000 dilution from a representative lot detected Tet2 in rat medulla tissue.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Tet2 in human H9 Embryonic Stem cells.

Quality

Evaluated by Western Blotting in mouse Embryonic Stem cells.

Western Blotting Analysis: 1 µg/mL of this antibody detected Tet2 in 10 µg of mouse Embryonic Stem cells (Note: Isoforms 2 and 3 may observed at low concentrations or longer exposure times, while isoform 1 may be detected at higher concentrations or longer exposure times in certain cell lysates.).

Target description

~140 kDa observed. Three isoforms at 223 kDa (isoform 1), 130 kDa (isoform 2), and 133 kDa (isoform 3) are known to exist. This antibody should recognize all three isoforms based on sequence homology. An uncharacterized band may be observed at ~36 kDa in some cell lysates. (Note: Isoforms 2 and 3 may observed at low concentrations, while isoform 1 may be detected at higher concentrations in certain cell lysates.)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiajie Tu et al.
Stem cell research & therapy, 9(1), 71-71 (2018-03-23)
The regulatory role of long noncoding RNAs (lncRNAs) have been partially proved in embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs). In the current study, we investigated mouse ESC (mESC) self-renewal, differentiation, and proliferation in vitro by knocking
Sherry X Yang et al.
Journal of hematology & oncology, 14(1), 83-83 (2021-05-28)
Challenges remain on the selection of patients who potentially respond to a class of drugs that target epigenetics for cancer treatment. This study aims to investigate TET2/DNMT3A mutations and antitumor activity of a novel epigenetic agent in multiple human cancer
Yunpeng Zhang et al.
Evidence-based complementary and alternative medicine : eCAM, 2019, 1282085-1282085 (2019-11-28)
In addition to abnormalities of motor and posture, children with cerebral palsy (CP) often have intellectual disability. As a complementary and alternative traditional Chinese medicine (TCM) therapy, Chinese Tuina massage, also called Tuina in China, has been widely applied in
Yun Liu et al.
Molecular pain, 18, 17448069221143671-17448069221143671 (2022-11-23)
DNA hydroxylation catalyzed by Tet dioxygenases occurs abundantly in neurons in mammals. However, effects of ten-eleven translocation methylcytosine dioxygenase 1 (TET1) expression and hydroxymethylation status on neuron injury remain unclear. This study was designed to explore the effects of TET1
Yunpeng Zhang et al.
Frontiers in molecular neuroscience, 12, 214-214 (2019-09-26)
Cerebral palsy (CP) is a neurodevelopmental disorder usually occurring early in life and persisting through the whole life. Several risk factors, including perinatal hypoxia-ischemia (HI), may contribute to occurrence of CP in preterm infants. DNA hydroxymethylation has been shown to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service