SAB3500711
Anti-TET2 antibody produced in rabbit
affinity isolated antibody
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
mol wt
220 kDa
species reactivity
human
concentration
1 mg/mL
technique(s)
ELISA: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... TET2(54790)
General description
Tet methylcytosine dioxygenase 2 (TET2) is part of the Fe(II)/α-ketoglutarate-dependent dioxygenase family of enzymes. It is expressed in the brain. The gene encoding TET2 is localized on human chromosome 4q24.
Immunogen
Antibody was raised against an 18 amino acid peptide near the amino terminus of human TET2.
Application
Anti-TET2 antibody produced in rabbit has been used in Western blotting.
Biochem/physiol Actions
Tet methylcytosine dioxygenase 2 (TET2) is involved in the conversion of 5-methylcytosine to 5-hydroxymethyl-cytosine (5hmC). It also has a role in stimulating DNA demethylation and lymphomagenesis. Mutations in the gene encoding TET2 have been linked to myelodysplastic syndromes.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Linkage
The action of this antibody can be blocked using blocking peptide SBP3500711.
Physical form
Supplied at 1 mg/mL in PBS with 0.02% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Vitamin C enhances epigenetic modifications induced by 5-azacytidine and cell cycle arrest in the hepatocellular carcinoma cell lines HLE and Huh7
Sahar O
Clinical epigenetics (2016)
Prognostic significance of TET2 mutations in myelodysplastic syndromes: A meta-analysis.
Guo Z
Leukemia Research (2017)
Sahar Olsadat Sajadian et al.
Clinical epigenetics, 8, 46-46 (2016-05-03)
5-Azacytidine (5-AZA), a DNA methyl transferase inhibitor, is a clinically used epigenetic drug for cancer therapy. Recently, we have shown that 5-AZA upregulates ten-eleven translocation (TET) protein expression in hepatocellular carcinoma (HCC) cells, which induce active demethylation. Vitamin C facilitates
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service