Skip to Content
Merck
All Photos(1)

Key Documents

IMPC-051-01

Supelco

Theophylline solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N4O2
CAS Number:
Molecular Weight:
180.16
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

CN1C(=O)N(C)c2nc[nH]c2C1=O

InChI

1S/C7H8N4O2/c1-10-5-4(8-3-9-5)6(12)11(2)7(10)13/h3H,1-2H3,(H,8,9)

InChI key

ZFXYFBGIUFBOJW-UHFFFAOYSA-N

General description

A Certified Spiking Solution® suitable for use as starting material in the preparation of calibrators and controls for theophylline testing methods by LC-MS/MS or GC-MS. Theophylline, also known as dimethylxanthine, is a naturally occurring stimulant of the xanthine family. Sold under the trade name Theolair, theophylline is used for treatment of asthma and other respiratory diseases.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria Boswell-Smith et al.
The Journal of allergy and clinical immunology, 117(6), 1237-1243 (2006-06-06)
Theophylline has been relegated to a second- or even third-line therapy in the treatment of asthma and chronic obstructive pulmonary disease (COPD), behind glucocorticosteroids and beta2-agonists, although recent findings have suggested that theophylline possesses anti-inflammatory and immunomodulatory effects in addition
David J Henderson-Smart et al.
The Cochrane database of systematic reviews, (1)(1), CD000273-CD000273 (2010-01-22)
Recurrent apnea is common in preterm infants, particularly at very early gestational ages. These episodes of loss of effective breathing can lead to hypoxemia and bradycardia, which may be severe enough to require resuscitation including use of positive pressure ventilation.
F S F Ram et al.
Respiratory medicine, 99(2), 135-144 (2005-02-18)
To determine the efficacy of oral theophylline compared with placebo in people with stable chronic obstructive pulmonary disease (COPD). Systematic review of randomized-controlled trials comparing oral theophylline with placebo for a minimum of 7 days in people with stable COPD.
Bing Dai et al.
American journal of kidney diseases : the official journal of the National Kidney Foundation, 60(3), 360-370 (2012-04-21)
Whether treatment with adenosine receptor antagonists such as theophylline can prevent contrast-induced acute kidney injury (AKI) remains controversial. We conducted a meta-analysis of randomized controlled trials using MEDLINE (1966 to July 2011), EMBASE (1980 to July 2011), Web of Science
Mazen Al-Alawi et al.
The American journal of medicine, 127(5), 370-378 (2014-01-02)
Global estimates on aging predict an increased burden of asthma in the older population. Consequently, its recognition, diagnosis, and management in clinical practice require optimization. This review aims to provide an update for clinicians, highlighting advances in the understanding of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service