Merck
Advances in the diagnosis and management of asthma in older adults.
The American journal of medicine (2014-01-02)
Mazen Al-Alawi, Tidi Hassan, Sanjay H Chotirmall
ABSTRACT
Global estimates on aging predict an increased burden of asthma in the older population. Consequently, its recognition, diagnosis, and management in clinical practice require optimization. This review aims to provide an update for clinicians, highlighting advances in the understanding of the aging process and immunosenescence together with their applicability to asthma from a diagnostic and therapeutic perspective. Aging impacts airway responses and immune function, and influences efficacy of emerging phenotype-specific therapies when applied to the elderly patient. Differentiating eosinophilic and neutrophilic disease accounts for atopic illness and distinguishes long-standing from late-onset asthma. Therapeutic challenges in drug delivery, treatment adherence, and side-effect profiles persist in the older patient, while novel recording devices developed to aid detection of an adequate inhalation evaluate treatment effectiveness and compliance more accurately than previously attainable. Anticytokine therapies improve control of brittle asthma, while bronchial thermoplasty is an option in refractory cases. Multidimensional intervention strategies prove best in the management of asthma in the older adult, which remains a condition that is not rare but rarely diagnosed in this patient population.
Product Description
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Theophylline, European Pharmacopoeia (EP) Reference Standard
USP
Theophylline, United States Pharmacopeia (USP) Reference Standard