Skip to Content
Merck
All Photos(1)

Key Documents

1653004

USP

Theophylline

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,3-Dimethylxanthine, 2,6-Dihydroxy-1,3-dimethylpurine, 3,7-Dihydro-1,3-dimethyl-1H-purine-2,6-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N4O2
CAS Number:
Molecular Weight:
180.16
Beilstein:
13463
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

theophylline

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1C(=O)N(C)c2[nH]cnc2C1=O

InChI

1S/C7H8N4O2/c1-10-5-4(8-3-9-5)6(12)11(2)7(10)13/h3H,1-2H3,(H,8,9)

InChI key

ZFXYFBGIUFBOJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Theophylline USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Aminophylline
  • Aminophylline Delayed-Release Tablets
  • Aminophylline Injection
  • Aminophylline Oral Solution
  • Aminophylline Rectal Solution
  • Aminophylline Tablets
  • Dimenhydrinate

Biochem/physiol Actions

Phosphodiesterase inhibitor; diuretic; cardiac stimulant; muscle relaxant; asthma medication.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria Boswell-Smith et al.
The Journal of allergy and clinical immunology, 117(6), 1237-1243 (2006-06-06)
Theophylline has been relegated to a second- or even third-line therapy in the treatment of asthma and chronic obstructive pulmonary disease (COPD), behind glucocorticosteroids and beta2-agonists, although recent findings have suggested that theophylline possesses anti-inflammatory and immunomodulatory effects in addition
Diego Caccavo et al.
Molecular pharmaceutics, 12(2), 474-483 (2014-12-17)
In this work the behavior of hydrogel-based matrices, the most widespread systems for oral controlled release of pharmaceuticals, has been mathematically described. In addition, the calculations of the model have been validated against a rich set of experimental data obtained
David J Henderson-Smart et al.
The Cochrane database of systematic reviews, (1)(1), CD000273-CD000273 (2010-01-22)
Recurrent apnea is common in preterm infants, particularly at very early gestational ages. These episodes of loss of effective breathing can lead to hypoxemia and bradycardia, which may be severe enough to require resuscitation including use of positive pressure ventilation.
Amy T Ma et al.
Applied and environmental microbiology, 80(21), 6704-6713 (2014-08-26)
Cyanobacteria are photosynthetic bacteria that are currently being developed as biological production platforms. They derive energy from light and carbon from atmospheric carbon dioxide, and some species can fix atmospheric nitrogen. One advantage of developing cyanobacteria for renewable production of
Laura López-Cruz et al.
Behavioural brain research, 270, 213-222 (2014-05-27)
Caffeine and theophylline are methylxanthines that are broadly consumed, sometimes at high doses, and act as minor psychostimulants. Both are nonselective adenosine antagonists for A1 and A2A receptors, which are colocalized with dopamine D1 and D2 receptors in striatal areas.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service