Skip to Content
Merck
All Photos(1)

Key Documents

T-013

Supelco

Theobromine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N4O2
CAS Number:
Molecular Weight:
180.16
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CN1C(=O)NC(=O)c2c1ncn2C

InChI

1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)9-7(13)11(5)2/h3H,1-2H3,(H,9,12,13)

InChI key

YAPQBXQYLJRXSA-UHFFFAOYSA-N

General description

Theobromine is a naturally occurring vasodilator, diuretic, and heart stimulant. Also known as xantheose, theobromine is prescribed for treatment of high blood pressure. This Certified Spiking Solution® is suitable for use as starting material in the preparation of calibrators and controls for theobromine testing methods by LC-MS/MS or GC-MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Michael A Grandner et al.
Appetite, 64, 71-80 (2013-01-24)
Short sleep duration is associated with weight gain and obesity, diabetes, cardiovascular disease, psychiatric illness, and performance deficits. Likewise, long sleep duration is also associated with poor physical and mental health. The role of a healthy diet in habitual sleep
Shibin Mohanan et al.
Gene, 519(1), 107-112 (2013-02-05)
There are numerous reports on intronic miRNAs from plants, most of which are involved in the regulation of unrelated genes. Some of the target genes are antagonistic to the host genes. Intronic miRNAs in animal systems, however, are known to
Betul Kargul et al.
Oral health & preventive dentistry, 10(3), 275-282 (2012-10-25)
The objectives of this in-vitro study were to investigate the effect of theobromine, which is the principle xanthine species in Theobroma cacao, at two concentrations on the surface hardness and topography of human enamel. Twenty-four freshly extracted human third molars
Bodo Trognitz et al.
PloS one, 8(1), e54079-e54079 (2013-01-26)
The sensory quality and the contents of quality-determining chemical compounds in unfermented and fermented cocoa from 100 cacao trees (individual genotypes) representing groups of nine genotype spectra (GG), grown at smallholder plantings in the municipality of Waslala, Nicaragua, were evaluated
João Rodrigo Santos et al.
Analytica chimica acta, 715, 57-63 (2012-01-17)
In this work, the coupling of a commercial monolithic column to a traditional low pressure FIA system is proposed for the analysis of theobromine, theophylline and caffeine in coffee brewed samples using UV detection. The parameters mobile phase composition, flow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service