Skip to Content
Merck
All Photos(1)

Key Documents

870412P

Avanti

16:0 DG Galloyl

Avanti Research - A Croda Brand 870412P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-galloyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O9
CAS Number:
Molecular Weight:
721.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (870412P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870412P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COC(C1=CC(O)=C(O)C(O)=C1)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C42H72O9/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-39(45)49-33-36(34-50-42(48)35-31-37(43)41(47)38(44)32-35)51-40(46)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h31-32,36,43-44,47H,3-30,33-34H2,1-2H3/t36-/m1/s1

InChI key

VUMJZTCVWBBKRP-PSXMRANNSA-N

Biochem/physiol Actions

16:0 DG Galloyl or 1,2-dipalmitoyl-sn-glycero-3-galloyl (DPGG) is a phenolic-based lipid molecule, which is used as a component of polydiacetylenes (PDA) liposomes. This lipid can form phenolic metal complexes with lead (II).

Packaging

5 mL Amber Glass Screw Cap Vial (870412P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janus particles for (bio) sensing
Yanez-Sedeno P, et al.
Applied Materials Today, 9, 276-288 (2017)
Versatile Surface Functionalization of Metal--Organic Frameworks through Direct Metal Coordination with a Phenolic Lipid Enables Diverse Applications
Zhu W, et al.
Advances in Functional Materials, 28(16), 1705274-1705274 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service