Skip to Content
Merck
All Photos(3)

Key Documents

840032P

Avanti

Brain PS

Avanti Research - A Croda Brand

Synonym(s):

L-α-phosphatidylserine (Brain, Porcine) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840032P-10mg)
pkg of 1 × 100 mg (840032P-100mg)
pkg of 5 × 100 mg (840032P-500mg)
pkg of 1 × 25 mg (840032P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C42H80NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(44)50-35-38(36-51-54(48,49)52-37-39(43)42(46)47)53-41(45)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h18,20,38-39H,3-17,19,21-37,43H2,1-2H3,(H,46,47)(H,48,49);/q;+1/p-1/b20-18-;/t38-,39+;/m1./s1

InChI key

ALPKKMIPHGSQRX-NJZWBUMZSA-M

Application

Brain PS has been used in lipid standard material preparation, reporter assay and in the preparation of liposomes.

Biochem/physiol Actions

Phosphatidylserine is a lipid. It helps to increase the cognitive performance of rats and humans, having different levels of age-dependent memory problems or probable Alzheimer disease. It has the ability to block degeneration of the cholinergic neuronal population of the basal forebrain of aged rats. 90% of the phosphatidylserine decarboxylase activity is linked with mitochondria.

Packaging

5 mL Clear Glass Sealed Ampule (840032P-100mg)
5 mL Clear Glass Sealed Ampule (840032P-10mg)
5 mL Clear Glass Sealed Ampule (840032P-25mg)
5 mL Clear Glass Sealed Ampule (840032P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of brain phosphatidylserine decarboxylase: localization in the mitochondrial inner membrane
Percy AK, et al.
Archives of Biochemistry and Biophysics, 223(2), 484-494 (1983)
Alzheimer's disease-associated TREM2 variants exhibit either decreased or increased ligand-dependent activation
Song W, et al.
Alzheimers Dement., 13(4), 381-387 (2017)
Bovine brain phosphatidylserine attenuates scopolamine induced amnesia in mice
Claro FT, et al.
Progress in Neuro-Psychopharmacology & Biological Psychiatry, 30(5), 881-886 (2006)
M Gangal et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(22), 12394-12399 (1999-10-27)
Although the catalytic (C) subunit of cAMP-dependent protein kinase is N-myristylated, it is a soluble protein, and no physiological role has been identified for its myristyl moiety. To determine whether the interaction of the two regulatory (R) subunit isoforms (R(I)
Molecular Medicines for Cancer: Concepts and Applications of Nanotechnology (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service