Skip to Content
Merck
All Photos(2)

Key Documents

810144P

Avanti

16:0 NBD PE

Avanti Research - A Croda Brand, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H78N5O11P
CAS Number:
Molecular Weight:
872.08
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810144P-1mg)
pkg of 5 × 1 mg (810144P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

16:0 NBD PE is a headgroup labelled fluorescent phospholipid. L-α-Phosphatidylethanolamine (PE) is the most abundant phospholipid in bacteria and the second most abundant in animals, plants and yeast.[1]

Application

16:0 NBD PE is suitable:
  • for use in the bilayer, as fluorescent probe, together with encapsulated doxorubicin (DXR)[2] for the preparation of planar-supported lipid bilayers (PSLB) of 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC)
  • used in fluorescence control experiment[3]
  • for labelling giant plasma membrane vesicles[4]

Biochem/physiol Actions

L-α-Phosphatidylethanolamine maintains membrane integrity.[5]

Packaging

5 mL Amber Glass Screw Cap Vial (810144P-1mg)
5 mL Amber Glass Screw Cap Vial (810144P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Mazur et al.
Langmuir : the ACS journal of surfaces and colloids (2020-02-26)
Synthetic micromotors are evaluated extensively in a range of biomedical, microscale transport, and environmental applications. Fundamental insight into micromotors that exhibit locomotion due to triggered disintegration of their associated liposomes is provided. Directed self-propulsion is observed when the lipid vesicles
1, 2-diacyl-phosphatidylcholine flip-flop measured directly by sum-frequency vibrational spectroscopy
Liu J and Conboy JC
Biophysical Journal, 89(4), 2522-2532 (2005)
Mesha Saeed et al.
International journal of nanomedicine, 14, 2069-2089 (2019-04-17)
Melanoma is the most aggressive form of skin cancer. Chemotherapy at a late stage fails due to low accumulation in tumors, indicating the need for targeted therapy. To increase drug uptake by tumor cells, we have targeted doxorubicin-containing liposomes using
Structural determinants for partitioning of lipids and proteins between coexisting fluid phases in giant plasma membrane vesicles
Sengupta P, et al.
Biochimica et Biophysica Acta - Biomembranes, 1778(1), 20-32 (2008)
Sindy Müller et al.
Pharmaceutics, 11(12) (2019-12-11)
The use of archaeal lipids and their artificial analogues, also known as bolalipids, represents a promising approach for the stabilization of classical lipid vesicles for oral application. In a previous study, we investigated the mixing behavior of three single-chain alkyl-branched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service