Skip to Content
Merck
All Photos(3)

Key Documents

840055P

Avanti

Liver PC

Avanti Research - A Croda Brand 840055P, powder

Synonym(s):

L-α-phosphatidylcholine (Liver, Bovine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840055P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840055P

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O)=O

General description

Phosphatidylcholine (PC) has two fatty acid chains attached to a head group. Generally, PC has saturated fatty acid in the sn-1 position and unsaturated fatty acid in the sn-2 position. It has a quaternary ammonium group and a phosphate moiety in the head group, existing as a zwitterion in a wide range of pH.

Application

Liver PC is suitable for use:
  • as a standard for quantitation of tumor derived lipids by thin layer chromatography
  • in the preparation of liposomes to mimic mammalian membranes
  • in lipid vesicle preparation to study intravesicular transfer of α-tocopherol fluorescent derivative (NBD-Toc)

Biochem/physiol Actions

Phosphatidylcholine (PC) plays a key role in the synthesis of second messenger and signaling molecules like diacyl glycerol, lysophosphatidic acid, phosphatidic acid and arachidonic acid. Liver PC has a saturated to unsaturated fatty acid ratio of 0.91.

Packaging

5 mL Clear Glass Sealed Ampule (840055P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noncovalently and covalently bound lipid on the receptor for immunoglobulin E
Kinet JP, et al.
Biochemistry, 24(25), 7342-7348 (1985)
Liposomes with diverse compositions are protected during desiccation by LEA proteins from Artemia franciscana and trehalose
Moore DS, et al.
Biochimica et Biophysica Acta - Biomembranes, 1858(1), 104-115 (2016)
Essentials of Medical Biochemistry: With Clinical Cases (2011)
Effect of bilayer phospholipid composition and curvature on ligand transfer by the alpha-tocopherol transfer protein
Zhang WX, et al.
Lipids, 44(7), 631-631 (2009)
The Kennedy pathway?de novo synthesis of phosphatidylethanolamine and phosphatidylcholine
Gibellini F and Smith TK
IUBMB Life, 62(6), 414-428 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service