Skip to Content
Merck
All Photos(2)

Key Documents

810130P

Avanti

16:0-06:0 NBD PC

Avanti Research - A Croda Brand 810130P, powder

Synonym(s):

1-palmitoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H62N5O11P
CAS Number:
Molecular Weight:
771.88
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810130P-1mg)
pkg of 5 × 1 mg (810130P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810130P

shipped in

dry ice

storage temp.

−20°C

General description

16:0-06:0 NBD PC (1-palmitoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphocholine) is a fluorescent analogue of phosphatidylcholine.

Application

16:0-06:0 NBD PC (1-palmitoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphocholine) has been used in NBD-phospholipid translocation assay. It may be used as a molecular fluorescent marker to label lipids during the friction tests, as substrate for enzyme substrate specificity determination and in the preparation of small unilamellar vesicles.

Biochem/physiol Actions

16:0-06:0 NBD PC (1-palmitoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphocholine) acts as a potential fluorescent probe. It is used for the quantification of the soluble and membrane-bound phospholipase A2 (PLA2) activity. 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) derivatives are widely used as fluorescent analogs of native lipids in biological and model membranes to study a variety of processes.

Packaging

5 mL Amber Glass Screw Cap Vial (810130P-1mg)
5 mL Amber Glass Screw Cap Vial (810130P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of fluorescent probe NBD-PC on the structure, dynamics and phase transition of DPPC. A molecular dynamics and differential scanning calorimetry study
Loura L, et al.
Biochimica et Biophysica Acta - Biomembranes, 1778(2), 491-501 (2008)
In vitro analysis of the Staphylococcus aureus lipoteichoic acid synthase enzyme using fluorescently labeled lipids
Karatsa-Dodgson MW, et al.
Journal of Bacteriology, 192(20), 5341-5349 (2010)
Role of the biomolecular interactions in the structure and tribological properties of synovial fluid
Mirea DA, et al.
Tribology International, 59(3), 302-311 (2013)
The use of C6-NBD-PC for assaying phospholipase A2-activity: scope and limitations
Meyuhas D, et al.
Biochimica et Biophysica Acta, Lipids and Lipid Metabolism, 1124(3), 223-232 (1992)
Measuring translocation of fluorescent lipid derivatives across yeast Golgi membranes
Natarajan P, et al.
Methods, 39(2), 163-168 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service